Skip to Content
MilliporeSigma

Skip To

1.99091

Buffer solution

traceable to NIST SRM, traceable to PTB, pH 9.18 (25 °C in H2O), Certipur®

Synonym(s):

di-sodium tetraborate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

traceable to NIST SRM, traceable to PTB

product line

Certipur®

pH

9.18 (25 °C in H2O)

bp

100 °C/1013 hPa

density

1.00 g/cm3 at 20 °C

storage temp.

15-25°C

General description

The di-sodium tetraborate buffer solution is gravimetrically prepared at a registered calibration laboratory with high-quality reagents and water. Its pH value of 9.20 at 25°C is measured using a combined glass electrode following a 5-point calibration method. The measurement is made following DIN 19268 using reference buffer solutions compliant to DIN 19266, NIST, IUPAC, Ph. Eur, and USP. The buffer pH is temperature dependent and therefore a constant temperature must be maintained during its use.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The buffer is used as a standard to calibrate pH measuring instruments or pH electrodes and as a control to measure the pH of other solutions.

Features and Benefits

  • Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to secondary reference standard from NIST and primary certified reference standard from PTB
  • Available in secure 500 mL plastic bottle to protect against contamination
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
  • Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
  • Packaging material subjected to comprehensive stability studies as per ISO Guide

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Buffer solution


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
199091050004027269966539

Questions

  1. What is the concentration of molar concentration of di-sodium tetraborate in the buffer?

    1 answer
    1. The molar concentration of di-sodium tetraborate in this buffer is proprietary. The certified pH value is available in the Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters