Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.25
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Quality Segment
agency
traceable to NIST, traceable to PTB
product line
Certipur®
composition
di-sodium hydrogen phosphate/potassium dihydrogen phosphate
pH
6.86 (25 °C)
storage temp.
15-25°C
General description
The potassium dihydrogen phosphate/di-sodium hydrogen phophate buffer solution is gravimetrically prepared at a registered calibration laboratory with high-quality reagents and water. Its pH value of 6.86 at 25°C is measured using a combined glass electrode following a 5-point calibration method. The measurement is made following DIN 19268 using reference buffer solutions compliant to DIN 19266, NIST, IUPAC, Ph. Eur, and USP. The buffer pH is temperature dependent and therefore a constant temperature must be maintained during its use.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
The buffer is used as a standard to calibrate pH measuring instruments or pH electrodes and as a control to measure the pH of other solutions.
Features and Benefits
- Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
- Traceable to secondary reference standard from NIST and primary certified reference standard from PTB
- Available in a secure 500 mL plastic bottle to prevent contamination
- Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
- Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
- Packaging material subjected to comprehensive stability studies as per ISO Guide 35
- An uncertainty of +/- 0.02 in the pH value at 25°C
Legal Information
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
This Item | |||
|---|---|---|---|
| product line Certipur® | product line Certipur® | product line - | product line Certipur® |
| agency traceable to NIST, traceable to PTB | agency - | agency - | agency - |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| composition di-sodium hydrogen phosphate/potassium dihydrogen phosphate | composition - | composition - | composition - |
| pH 6.86 (25 °C) | pH 8.0 (25 °C in H2O) | pH 1.7 (25 °C in H2O) | pH 7.0 (25 °C in H2O) |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C |
Still not finding the right product?
Explore all of our products under Buffer solution
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



