Skip to Content
MilliporeSigma

Skip To

118501

ATM/ATR Kinase Inhibitor

The ATM/ATR Kinase Inhibitor, also referenced under CAS 905973-89-9, controls the biological activity of ATM/ATR Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

ATM/ATR Kinase Inhibitor, CGK 733

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C23H18Cl3FN4O3S
CAS Number:
Molecular Weight:
555.84
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

pale yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c(cc(cc1)NC(=S)NC(NC(=O)C(c3ccccc3)c2ccccc2)C(Cl)(Cl)Cl)[N+](=O)[O-]

InChI

1S/C23H18Cl3FN4O3S/c24-23(25,26)21(30-22(35)28-16-11-12-17(27)18(13-16)31(33)34)29-20(32)19(14-7-3-1-4-8-14)15-9-5-2-6-10-15/h1-13,19,21H,(H,29,32)(H2,28,30,35)

InChI key

HLCDNLNLQNYZTK-UHFFFAOYSA-N

General description

A cell-permeable thiourea compound that selectively inhibits the kinase activity of ATM and ATR (IC50 ~ 200 nM) without significant effect against other PIKK family members (PI3K, mTOR, and DNA-PK) or kinases that are known to phosphorylate p53 (ERK, CK1, Cdk2, JNK, PKC, CAK, Chk1, PKR, and p38). Shown to reverse senescence in human primary BJ and HMEC cells. Unlike LY 294002 (Cat. No. 440202 and 440204), this inhibitor does not interfere with cellular PI3K-PDK1-Akt signaling pathway.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ATM/ATR
Product does not compete with ATP.
Reversible: no
Target IC50: ~200 nM against ATM and ATR

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Goldstein, M., et al. 2008. Toxicol. Appl. Pharmacol.in press.
Crescenzi, E., et al. 2008. Clin. Cancer Res.14, 1877.
Cruet-Hennequart, S., et al. 2008. DNA Repair7, 582.
Won, J., et al. 2006. Nat. Chem. Biol.2, 369. (RETRACTED)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
361548118510616452
assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥97% (HPLC)

assay

≥95% (HPLC)

form

solid

form

solid

form

powder

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 5 mg/mL

solubility

DMSO: 5 mg/mL

solubility

DMSO: 100 mg/mL

solubility

DMSO: 5 mg/mL, methanol: 5 mg/mL


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
118501-5MG04055977224054

Questions

Reviews

No rating value

Active Filters