Skip to Content
MilliporeSigma

Skip To

1.16755

Cesium chloride-lanthanum chloride buffersolution

acc. to Schinkel for atomic absorption spectroscopy 10 g/l CsCl + 100 g/l La

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.21
UNSPSC Code:
12161703
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

for inorganic trace analysis

form

liquid

storage condition

(No metal or light-weight-metal containers. No metal containers. Tightly closed.)

concentration

(Lanthanum(III) chloride: >= 5 - < 10%

Cesium chloride: >= 0.1 - < 1%)

technique(s)

AAS: suitable

color

violet

pH

<1 (25 °C in H2O, undiluted)

solubility

water: soluble

density

1.178 g/cm3 at 20 °C

cation traces

Ca: ≤5.0 mg/L, Cd: ≤0.2 mg/L, Co: ≤1.0 mg/L, Cr: ≤1.0 mg/L, Cs: : 7.50-8.28 g/L, Cu: ≤0.2 mg/L, Fe: ≤1.0 mg/L, K: ≤0.2 mg/L, Li: ≤0.2 mg/L, Mg: ≤1.0 mg/L, Mn: ≤0.2 mg/L, Na: ≤2.0 mg/L, Ni: ≤1.0 mg/L, Sr: ≤1.0 mg/L, Zn: ≤2.0 mg/L

storage temp.

15-25°C

Analysis Note

Assay Lanthan (potentiometric): 95.2 - 105.2 g/l
Cs (Cesium): 7.50 - 8.28 g/l
Ca (Calcium): ≤ 5.0 mg/l
Cd (Cadmium): ≤ 0.2 mg/l
Co (Cobalt): ≤ 1.0 mg/l
Cr (Chromium): ≤ 1.0 mg/l
Cu (Copper): ≤ 0.2 mg/l
Fe (Iron): ≤ 1.0 mg/l
potassium: ≤ 1.0 mg/l
Li (Lithium): ≤ 0.2 mg/l
Mg (Magnesium): ≤ 1.0 mg/l
Mn (Manganese): ≤ 0.2 mg/l
Na (Sodium): ≤ 2.0 mg/l
Ni (Nickel): ≤ 1.0 mg/l
Sr (Strontium): ≤ 1.0 mg/l
Zn (Zinc): ≤ 2.0 mg/l


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters