Skip to Content
MilliporeSigma

Skip To

1.09441

Buffer solution

(glycine/sodium chloride/hydrogen chloride), traceable to SRM from NIST and PTB pH 1.00 (25°C) Certipur®

Synonym(s):

Buffer solution pH 1.00 (25°C) (glycine/sodium chloride/hydrogen chloride)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12161703
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

product line

Certipur®

pH

1.0 (25 °C in H2O)

density

1.00 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The buffer solution (glycine/sodium chloride/hydrogen chloride) is gravimetrically prepared using high-quality reagents. It is quality controlled and qualified in an ISO/IEC 17025 accredited calibration lab. Its pH value of 1.00 at 25°C is qualified using a combined glass electrode following a 5-point calibration method, in accordance with DIN 19268, against certified primary reference standards as per DIN 19266.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The buffer solution is used in analytical procedures for the calibration, qualification, and monitoring of pH measuring instruments.

Features and Benefits

  • Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to SRMs from NIST and primary certified reference standards from PTB
  • Ready-to-use solution
  • Available in a sealed bottle and the innovative Titripac® packaging for positive environmental impact due to reduced waste generation
  • Long-lasting with a shelf life of 3 years
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.094321.094391.09435
format

mixture

format

mixture

format

mixture

format

mixture

product line

Certipur®

product line

Certipur®

product line

Certipur®

product line

Certipur®

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

pH

1.0 (25 °C in H2O)

pH

1.0 (25 °C in H2O)

pH

7.0 (20 °C in H2O)

pH

4.0 (20 °C in H2O)


Still not finding the right product?

Explore all of our products under Buffer solution


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
109441050004027269386726
109441400004027269386757

Questions

Reviews

No rating value

Active Filters