Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C11H13N3O
CAS Number:
Molecular Weight:
203.24
UNSPSC Code:
12352115
EC Index Number:
201-452-3
NACRES:
NA.05
MDL number:
Pricing and availability is not currently available.
Quality Level
form
solid
potency
1700 mg/kg LD50, oral (Rat)
pH
7.1 (20 °C, 100 g/L in H2O, slurry)
mp
107-109 °C
solubility
56 g/L
bulk density
430 kg/m3
storage temp.
15-25°C
SMILES string
[n]2([n](c(c([c]2=O)N)C)C)c1ccccc1
InChI
1S/C11H13N3O/c1-8-10(12)11(15)14(13(8)2)9-6-4-3-5-7-9/h3-7H,12H2,1-2H3
InChI key
RLFWWDJHLFCNIJ-UHFFFAOYSA-N
Analysis Note
Assay (perchloric acid titration): ≥ 99
Identity (IR-spectrum): passes test
Melting point: 107 - 109
Sulfated ash: ≤ 0.1 %
Loss on drying (90 °C): ≤ 0.5 %
Sensitivity test (to phenol): passes test
Identity (IR-spectrum): passes test
Melting point: 107 - 109
Sulfated ash: ≤ 0.1 %
Loss on drying (90 °C): ≤ 0.5 %
Sensitivity test (to phenol): passes test
1 of 1
This Item | |||
|---|---|---|---|
| solubility 56 g/L | solubility - | solubility - | solubility - |
| Quality Level 100 | Quality Level 100 | Quality Level 200 | Quality Level - |
| form solid | form - | form - | form - |
| storage temp. 15-25°C | storage temp. - | storage temp. - | storage temp. - |
| mp 107-109 °C | mp 105-110 °C (lit.) | mp 105-110 °C (lit.), 107-109 °C | mp 105-110 °C (lit.) |
| bulk density 430 kg/m3 | bulk density - | bulk density - | bulk density - |
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1072930100 | 04022536089416 |
| 1072930010 | 04022536089409 |


