Skip to Content
MilliporeSigma

Skip To

1.06264

Sodium acetate

anhydrous 99.99 Suprapur®

Synonym(s):

Sodium acetate, Acetic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C2H3NaO2
CAS Number:
Molecular Weight:
82.03
UNSPSC Code:
12352107
EC Index Number:
204-823-8
NACRES:
NA.21
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

for inorganic trace analysis

assay

99.99%

form

powder

autoignition temp.

607 °C

potency

3530 mg/kg LD50, oral (Rat), >10000 mg/kg LD50, skin (Rabbit)

pH

7.5-9.2 (20 °C, 30 g/L in H2O)

bp

>400 °C (decomposition)

mp

324 °C (decomposition)

transition temp

flash point >250 °C

solubility

365 g/L

density

1.52 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Na+].[O-]C(=O)C

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Application


  • Thermophysical Studies: Sodium acetate buffers are used in thermophysical studies to investigate the behavior of protonated and deprotonated glycine in aqueous solutions at various temperatures. This research provides insights into the interactions and stability of biochemical compounds, which are critical for understanding biological processes and developing new analytical methods in biochemistry (Indian Journal of Chemistry - Section A, 2021).

Analysis Note

Purity (metallic): ≥ 99.99 %
Assay (perchloric acid titration): ≥ 99 %
Chloride (Cl): ≤ 5 ppm
Phosphate (PO₄): ≤ 5 ppm
Sulfate (SO₄): ≤ 20 ppm
Al (Aluminium): ≤ 0.05 ppm
Ba (Barium): ≤ 5.0 ppm
Bi (Bismuth): ≤ 0.01 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.005 ppm
Ce (Cerium): ≤ 0.005 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.010 ppm
Cs (Cesium): ≤ 2.0 ppm
Cu (Copper): ≤ 0.005 ppm
Fe (Iron): ≤ 0.05 ppm
K (Potassium): ≤ 5 ppm
La (Lanthanum): ≤ 0.005 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.050 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.005 ppm
Rb (Rubidium): ≤ 0.5 ppm
Sc (Scandium): ≤ 0.005 ppm
Sm (Samarium): ≤ 0.005 ppm
Sr (Strontium): ≤ 0.10 ppm
Tl (Thallium): ≤ 0.005 ppm
Y (Yttrium): ≤ 0.005 ppm
Zn (Zinc): ≤ 0.010 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W3024757510-OP1.06268
assay

99.99%

assay

≥99%

assay

-

assay

≥99.0% (perchloric acid titration)

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

300

form

powder

form

solid

form

crystals

form

powder

pH

7.5-9.2 (20 °C, 30 g/L in H2O)

pH

-

pH

-

pH

7.0-9.2 (25 °C, 5% in H2O)

density

1.52 g/cm3 at 20 °C

density

-

density

-

density

1.52 g/cm3 at 20 °C

solubility

365 g/L

solubility

-

solubility

-

solubility

365 g/L


Still not finding the right product?

Explore all of our products under Sodium acetate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
106264050004022536074122
106264005004022536074115

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters