Skip to Content
MilliporeSigma

Skip To

1.04162

Sodium acetate trihydrate

granulated, EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP

Synonym(s):

Sodium acetate trihydrate, Acetic acid sodium salt trihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
UNSPSC Code:
12352302
Beilstein/REAXYS Number:
3732037
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, ChP, JP, Ph. Eur., USP

product line

EMPROVE® EXPERT

form

granular

autoignition temp.

1112 °F

impurities

low in endotoxins

application(s)

liquid formulation
parenterals
pharma/biopharma processes

storage temp.

15-25°C

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

General description

Standard bulk materials often show a tendency for caking, clumping and dust formation which results in laborious weighing and manual de-caking processes with a negative impact on manufacturing efficiency. Due to its granulated form, Sodium acetate granulated is free-flowing and easy to handle. Its greatly reduced caking behavior facilitates processability, speeds up manufacturing processes and increases operator safety.

As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium acetate trihydrate is used as a buffer component for Biomolecule downstream processing steps and final formulation as well as small molecule final formulation.

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.282051.062651.28204
form

granular

form

solid

form

solid

form

crystalline

impurities

low in endotoxins

impurities

endotoxins, low

impurities

-

impurities

-

application(s)

liquid formulation
parenterals
pharma/biopharma processes

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

application(s)

liquid formulation
ophthalmics
pharmaceutical
solid formulation

application(s)

pharmaceutical

product line

EMPROVE® EXPERT

product line

EMPROVE® EXPERT

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® API

agency

BP, JP, USP, ChP, Ph. Eur.

agency

BP, ChP, JP, Ph. Eur., USP

agency

BP, JP, Ph. Eur., USP

agency

BP, JP, Ph. Eur., USP

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

600


Still not finding the right product?

Explore all of our products under Sodium acetate trihydrate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content





Global Trade Item Number

SKUGTIN
104162902604054839620997
104162000104054839587399
104162100004054839604027
104162950004054839621017