Skip to Content
MilliporeSigma

Skip To

1.03753

Yeast Extract for Biotechnology and Microbiology

Granulated, for Biotechnology and Microbiology

Synonym(s):

Yeast extract granulated,

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41106212
EC Index Number:
232-387-9
NACRES:
NA.74
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Yeast extract, suitable for microbiology, medium granules

biological source

yeast

Quality Segment

form

medium granules

autoignition temp.

>200 °C

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 25 kg, pkg of 5 kg, pkg of 500 g

pH

7.0 (20 °C, 10 g/L in H2O)

solubility

410 g/L

density

0.5 g/cm3 at 20 °C

bulk density

490 kg/m3

application(s)

microbiology

storage temp.

15-25°C

General description

Yeast extract granulated is manufactured from selected strains of Saccharomyces cerevisiae. It is manufactured under controlled conditions to retain its vitamin content and other nutritive values, such as peptides, free amino acids, nucleic acids, minerals, trace elements, and phosphorous compounds. It is rich in vitamins and is often used to supply these factors in culture media at a concentration of 0.3 - 0.5%. Yeast extract provides a complete source of nutrients for microorganisms and is particularly useful for the cultivation of fastidious bacteria that require specific nutrients or additional growth factors. It can be used alone or in combination with other sources of nutrients, such as peptones or salts.

Application

Yeast extract is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts and molds from various pharmaceutical, environmental, food and beverage samples.

Features and Benefits

Our Granulated media are:
  • Safer - Significant reduction of inhalation of toxic and allergenic substances resulting in fewer allergic responses. Less contamination of working tools and environment
  • Accurate - No separation of components and no clumping, even under humid or warm conditions, ensure a uniform product with higher performance accuracy
  • Fast - Rapid and uniform dissolution in water
  • Easier - Easier handling and weighing due to the better flow properties. Media do not stick to the flask or container
  • Reliable - Homogenous distribution of ingredients guarantees high reproducibility even if only small amounts of media are used.

Analysis Note

Appearance (colour)brownish-yellowAppearance (description)granulesIdentityconformspH-value (2%; water)5.5 - 7.2N (Nitrogen) (calculated on dried substance)≥ 10.5%Sulfated ash (600 °C)≤ 17.0%Heavy metals (as Pb)≤ 0.005%Loss on drying (50 °C; <670 Pa; 3 h)≤ 5.0%Ca (Calcium)≤ 0.05%Mg (Magnesium)≤ 0.10%Chloride (Cl) (as NaCl)≤ 5.0%Phosphorous compounds (as P)≤ 2.5%Suitability for microbiologypasses test

Other Notes

Replaces: 1.03753

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
701611.13885Y0875
form

medium granules

form

powder

form

solid

form

powder

application(s)

microbiology

application(s)

environmental
food and beverages
microbiology

application(s)

microbiology

application(s)

microbiology

biological source

yeast

biological source

Saccharomyces cerevisiae

biological source

yeast

biological source

-

solubility

410 g/L

solubility

H2O: 2%, clear, yellow

solubility

410 g/L

solubility

H2O: 100 mg/mL, clear to slightly hazy, yellow to brownish-yellow

storage temp.

15-25°C

storage temp.

2-25°C

storage temp.

2-30°C

storage temp.

room temp

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Yeast extract


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Questions

Reviews

No rating value

Active Filters