Skip to Content
MilliporeSigma

Skip To

1.01207

di-Ammonium hydrogen phosphate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

di-Ammonium hydrogen phosphate, Ammonium biphosphate, Ammonium phosphate dibasic, Diammonium hydrogen phosphate, Fyrex

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
UNSPSC Code:
12352301
EC Index Number:
231-987-8
NACRES:
NA.21
MDL number:
Assay:
≥99.0% (acidimetric)
Grade:
ACS reagent
Form:
solid
Solubility:
690 g/L
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. Ph. Eur.

product line

EMSURE®

assay

≥99.0% (acidimetric)

form

solid

potency

6500 mg/kg LD50, oral (Rat), >7950 mg/kg LD50, skin (Rabbit)

impurities

≤0.005% Insoluble matter

pH

7.6-8.2 (20 °C, 100 g/L in H2O)

solubility

690 g/L

density

1.619 g/cm3 at 20 °C

bulk density

800‑1000 kg/m3

anion traces

chloride (Cl-): ≤0.0005%, nitrate (NO3-): ≤0.001%, sulfate (SO42-): ≤0.004%

cation traces

Ca: ≤0.001%, Fe: ≤0.001%, K: ≤0.001%, Mg: ≤0.0005%, Na: ≤0.001%, heavy metals: ≤0.0005% (ACS)

storage temp.

2-30°C

SMILES string

[P](=O)([O-])([O-])[O-].[N+H4].[N+H4].[H+]

InChI

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Application


  • Phase evolution and sintering kinetics of hydroxyapatite synthesized by solution combustion technique.: This study discusses the phase evolution and sintering behavior of hydroxyapatite, a critical material in bioceramics, emphasizing the chemical processes involved during synthesis by solution combustion, which might include the usage of di-Ammonium hydrogen phosphate as a precursor (Pratihar et al., 2006).


  • An investigation of the chemical synthesis and high-temperature sintering behaviour of calcium hydroxyapatite (HA) and tricalcium phosphate (TCP) bioceramics.: This article investigates the synthesis routes for bioceramics like hydroxyapatite and tricalcium phosphate, potentially utilizing di-Ammonium hydrogen phosphate in their production processes, focusing on their behavior under high temperatures (Taş et al., 1997).


Analysis Note

Assay (acidimetric): ≥ 99.0 %

Insoluble matter: ≤ 0.005 %

pH-value (5 %; water; 25 °C): 7.8 - 8.1

pH-value (20 %; water): about 8

Chloride (Cl): ≤ 0.0005 %

Nitrate (NO₃): ≤ 0.001%

Sulfate: ≤ 0.004 %

heavy metals (ACS)<= 0.0005 %

Ca (Calcium): ≤ 0.001 %

Fe (Iron): ≤ 0.001 %

K (Potassium): ≤ 0.001 %

Mg (Magnesium): ≤ 0.0005

%Na (Sodium): ≤ 0.001 %

Corresponds to ACS, Reag. Ph Eur

Other Notes

Replaces: AX1355-1; AX1355

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2159967976230402
form

solid

form

crystals or chunks, powder

form

crystals

form

powder, crystals or granules

assay

≥99.0% (acidimetric)

assay

≥98%

assay

99.0-101.0% (T)

assay

≥98% (alkalimetric)

solubility

690 g/L

solubility

-

solubility

H2O: 50 mg/mL, clear, colorless

solubility

-

Quality Level

300

Quality Level

200

Quality Level

100

Quality Level

100

grade

ACS reagent

grade

ACS reagent

grade

-

grade

ACS reagent, puriss. p.a.

cation traces

Ca: ≤0.001%, K: ≤0.001%, Na: ≤0.001%, Fe: ≤0.001%, heavy metals: ≤0.0005% (ACS), Mg: ≤0.0005%

cation traces

Ca: 0.001%, Fe: ≤0.001%, K: ≤0.005%, Mg: 0.0005%, Na: ≤0.005%, heavy metals (as Pb): ≤0.001%

cation traces

-

cation traces

Ca: ≤0.001%, Fe: ≤0.001%, K: ≤0.005%, Mg: ≤0.0005%, Na: ≤0.005%


Still not finding the right product?

Explore all of our products under di-Ammonium hydrogen phosphate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Food and Beverage Campaign - Constituents and Additives - Organic Acids

Food and Beverage Campaign - Constituents and Additives - Preservatives

View All Related Content




Global Trade Item Number

SKUGTIN
101207902504027269138561
101207050004022536017891
101207905004022536017914

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is original manufacturer information available for product number 1.01207? I don't see a dossier for it on the website.

    1 answer
    1. Product No. 1.01207 (di-Ammonium hydrogen phosphate, for analysis EMSURE® ACS,Reag. Ph Eur) is manufactured by Merck KGaA, Darmstadt, Germany under the Supelco® analytical products portfolio.

      Additional information on EMSURE® Inorganics and Solvents can be found under the "More Documents" section of the product details page for 1.01207: https://www.sigmaaldrich.com/product/mm/101207#product-documentation

      Helpful?

Reviews

No rating value

Active Filters