Skip to Content
MilliporeSigma

Skip To

1.00986

Ethanol

greener alternative

absolute, EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP

Synonym(s):

Ethyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Index Number:
200-578-6
UNSPSC Code:
12191502
Beilstein/REAXYS Number:
1718733
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

agency

BP, ChP, JP, Ph. Eur., USP

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C), 59 hPa ( 20 °C)

product line

EMPROVE® EXPERT

assay

≥99.2% (m/m)

form

liquid

autoignition temp.

683 °F

expl. lim.

19 %, 60 °F

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

≥99.5 % (v/v)

technique(s)

API processing | polymorph screening: suitable

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L in H2O)

bp

78 °C (lit.)

mp

−114 °C (lit.)

transition temp

flash point 12 °C

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation

format

neat

greener alternative category

storage temp.

2-25°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a bio-based ethanol and is aligned with the 5th principle of Green Chemistry "Safer Solvents and Auxiliaries" and 7th principle "Use of Renewable Feedstock". Click here for more information.

Application

Ethanol is used as a solvent in oral, topical, and parenteral formulations.
Due to its low microbial and endotoxin limits, Ethanol absolute Emprove® Expert is especially suitable for high-risk applications.

Other Notes

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.009711.009671.00983
concentration

≥99.5 % (v/v)

concentration

95.1-96.9 % (v/v), 96%

concentration

92.3-93.8 % (v/v) (d15°C/15°C), 95.1-96.9 % (v/v) (d15.56°C/15.56°C), 95.1-96.9 % (v/v) (d20°C/4°C), 96%

concentration

-

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

agency

BP, JP, USP, ChP, Ph. Eur.

agency

BP, ChP, Ph. Eur.

agency

ChP, JP, Ph. Eur., USP

agency

reag. ISO, reag. Ph. Eur.

technique(s)

API processing | polymorph screening: suitable

technique(s)

-

technique(s)

-

technique(s)

-

form

liquid

form

liquid

form

liquid

form

liquid

assay

≥99.2% (m/m)

assay

92.6-95.2% (m/m)

assay

92.3-93.8% (d15.56°C/15.56°C; m/m), 92.6-95.2% (d20°C/4°C; m/m)

assay

≥99.9% (GC)


Still not finding the right product?

Explore all of our products under Ethanol


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
100986992004027269020309
100986051104027269043865
100986100004022536013374
100986601004022536013480
100986250004022536013428
100986995004022536752310
100986250104065266024623
100986903004054839504464
100986602504022536013497
100986250804027269302900
100986918004022536013503
100986991004054839993299

Questions

Reviews

No rating value

Active Filters