Skip to Content
MilliporeSigma

Skip To

1.00949

Ethylene glycol

EMPLURA®

Synonym(s):

Ethylene glycol, Glycol, 1,2-Ethanediol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C2H6O2
CAS Number:
Molecular Weight:
62.07
UNSPSC Code:
12191502
EC Index Number:
203-473-3
NACRES:
NA.05
MDL number:
Assay:
≥99% (GC)
Bp:
197.6 °C/1013 hPa
Vapor pressure:
0.053 hPa ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.053 hPa ( 20 °C)

Quality Segment

product line

EMPLURA®

assay

≥99% (GC)

form

liquid

autoignition temp.

410 °C

expl. lim.

3.2-15.3 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.0002% Chloride (Cl), ≤0.0002% Fe (Iron), ≤0.3% Water

pH

6-7.5 (20 °C, 100 g/L in H2O)

bp

197.6 °C/1013 hPa

mp

-14--10 °C

transition temp

flash point 111 °C

solubility

1000 g/L

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 1.112 - 1.114
Chloride (Cl): ≤ 0.0002 %
Fe (Iron): ≤ 0.0002 %
Water: ≤ 0.3 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.94604EX0565EX0564
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

assay

≥99% (GC)

assay

-

assay

≥98% (GC)

assay

≥99% (GC)

solubility

1000 g/L

solubility

-

solubility

-

solubility

1000 g/L

form

liquid

form

-

form

liquid

form

liquid

expl. lim.

3.2-15.3 % (v/v)

expl. lim.

-

expl. lim.

-

expl. lim.

-

bp

197.6 °C/1013 hPa

bp

-

bp

197.6 °C/1013 hPa

bp

197.6 °C/1013 hPa


Still not finding the right product?

Explore all of our products under Ethylene glycol


pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

target_organs

Kidney

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

239.0 °F - open cup

flash_point_c

115 °C - open cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
100949250004022536012247
100949902704061838824592
100949050104061838824547
100949252104061838824578
100949052104061838824554
100949902604061838824585
100949100004022536012230
100949250104061838824561

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters