Skip to Content
MilliporeSigma

Skip To

1.00795

Hexane

≥98% (GC), suitable for GC/MS, SupraSolv®

Synonym(s):

Hexane, n-Hexane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
UNSPSC Code:
12191502
EC Index Number:
203-777-6
NACRES:
NA.03
Beilstein/REAXYS Number:
1730733
MDL number:
Assay:
≥98% (GC)
Technique(s):
GC/MS: suitable
Bp:
69 °C (lit.)
Vapor pressure:
256 mmHg ( 37.7 °C), 5.2 psi ( 37.7 °C), ~132 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

n-Hexane, for gas chromatography MS SupraSolv®

vapor density

~3 (vs air)

Quality Segment

vapor pressure

256 mmHg ( 37.7 °C), 5.2 psi ( 37.7 °C), ~132 mmHg ( 20 °C)

product line

SupraSolv®

assay

≥98% (GC)

form

liquid

autoignition temp.

453 °F

potency

25000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7.7 %

technique(s)

GC/MS: suitable

refractive index

n20/D 1.375 (lit.)

kinematic viscosity

0.50 cSt(20 °C)

bp

69 °C (lit.)

mp

−95 °C (lit.)

transition temp

flash point -22 °C

solubility

0.0095 g/L

density

0.659 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

General description

SupraSolv® MS is dedicated for use in gas chromatography coupled with mass spectrometry. SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimum signal-to-noise ratio leading to reliable and reproducible analysis results.

Application

Intended for residue analysis.
Recommended for the analyses of:
  • polychlorinated dibenzodioxins and polychlorinated dibenzofurans (PCDD/F) in food and water samples
  • polycyclic aromatic hydrocarbons (PAHs) in food

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.043711.070231.09687
technique(s)

GC/MS: suitable

technique(s)

gas chromatography (GC): suitable

technique(s)

-

technique(s)

gas chromatography (GC): suitable

assay

≥98% (GC)

assay

≥98% (GC)

assay

≥98.5% (Σ hexane isomers + methylcyclopentane )

assay

≥99.7% (GC)

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

solubility

0.0095 g/L

solubility

0.0095 g/L

solubility

-

solubility

0.0095 g/L

form

liquid

form

liquid

form

liquid

form

liquid

expl. lim.

7.7 %

expl. lim.

7.7 %

expl. lim.

7.7 %

expl. lim.

7.7 %


Still not finding the right product?

Explore all of our products under n-Hexane


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

-7.6 °F - closed cup

flash_point_c

-22 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters