Skip to Content
MilliporeSigma

Skip To

V-040

Retinyl acetate solution

100 μg/mL in ethanol (with 0.1% (w/v) BHT; degassed), reference material, Cerilliant®

Synonym(s):

Vitamin A acetate solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in ethanol (with 0.1% (w/v) BHT; degassed)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O(C\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(CCCC1(C)C)C)C(=O)C

InChI

1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9+,18-14+

InChI key

QGNJRVVDBSJHIZ-QHLGVNSISA-N

General description

Retinyl acetate, a natural preform of vitamin A, is classified as generally recognized as safe in the amounts used to fortify foods with vitamin A and is especially recommended for maternal supplementation during pregnancy.

Application

This certified solution standard is suitable for use as an internal standard in chromatographic or MS-based methods in clinical testing applications such as assessment of vitamin A deficiency or in food and nutrition testing applications of vitamin A supplements and fortified foods.
Retinyl acetate solution may be used as a certified reference material (CRM) for the determination of the analyte in biological samples, dairy foods and pharmaceutical formulations by chromatography techniques.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R788246958R4632
grade

reference material

grade

-

grade

analytical standard

grade

-

format

single component solution

format

-

format

neat

format

-

concentration

100 μg/mL in ethanol (with 0.1% (w/v) BHT; degassed)

concentration

-

concentration

-

concentration

-

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

manufacturer/tradename

Cerilliant®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

storage temp.

−70°C

storage temp.

−20°C

storage temp.

-10 to -25°C

storage temp.

−20°C


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

57.2 °F - closed cup

flash_point_c

14.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Determination of retinoids in human serum, tocopherol and retinyl acetate in pharmaceuticals by RP-LC with electrochemical detection
Wang LH and Wang JF
Journal of Pharmaceutical and Biomedical Analysis, 25(5-6), 785-793 (2001)
Simultaneous determination of ?-tocopherol and retinol in plasma or red cells by high pressure liquid chromatography
Bieri JG, et al.
American Journal of Clinical Nutrition, 32(10), 2143-2149 (1979)
Simultaneous liquid chromatographic determination of vitamins A, E and ?-carotene in common dairy foods
Hewavitharana AK, et al.
International dairy journal, 6(6), 613-624 (1996)



Global Trade Item Number

SKUGTIN
V-040-1ML04061837428784

Questions

Reviews

No rating value

Active Filters