Skip to Content
MilliporeSigma

Skip To

V-021

(+)-γ−Tocopherol (Vitamin E) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(+)-γ-Tocopherol, (R,R,R)-γ-Tocopherol, 7,8-Dimethyltocol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C28H48O2
CAS Number:
Molecular Weight:
416.68
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
200-659-6
Beilstein/REAXYS Number:
93072
MDL number:
eCl@ss:
34058016
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2cc(O)c(C)c(C)c2O1

InChI

1S/C28H48O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-19-26(29)23(5)24(6)27(25)30-28/h19-22,29H,8-18H2,1-7H3/t21-,22-,28-/m1/s1

InChI key

QUEDXNHFTDJVIY-DQCZWYHMSA-N

General description

A lipid soluble form of Vitamin E, gamma-Tocopherol is important to human health as an antioxidant and food additive. Vitamin E deficiency, often the result of poor nutrition or low absorption of dietary fat, is monitored in patient serum samples by LC-MS/MS or HPLC.

Application

This Certified Spiking Solution® is suitable for use as a starting material in the preparation of linearity standards, calibrators, and controls in a variety of testing applications from diagnostic testing of Vitamin E deficiency by LC-MS/MS or HPLC to quality control testing of tocopherol-based dietary supplements or pharmaceutical preparations.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V-02047785258024
application(s)

clinical testing

application(s)

clinical testing

application(s)

food and beverages

application(s)

cell analysis

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable

form

liquid

form

liquid

form

solid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

100

Quality Level

200

grade

certified reference material

grade

certified reference material

grade

analytical standard

grade

-

storage temp.

−20°C

storage temp.

−70°C

storage temp.

-10 to -25°C

storage temp.

2-8°C


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
V-021-1ML04061837428609

Questions

Reviews

No rating value

Active Filters