Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Level
form
liquid
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
500 μg/mL in methanol
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
mp
-97.99 °C
application(s)
clinical testing
format
single component solution
storage temp.
−20°C
SMILES string
O1[C@@](CCc2c1c(c(c(c2C([2H])([2H])[2H])O)C([2H])([2H])[2H])C)(CCC[C@@H](CCC[C@@H](CCCC(C)C)C)C)C
InChI
1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21-,22-,29-/m1/s1/i5D3,7D3
InChI key
GVJHHUAWPYXKBD-BCHQTGRQSA-N
General description
Snap-N-Shoot®
Application
The certified reference material (CRM) in solution can also be used as an internal standard as follows:
- High-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) based combined analysis of retinol, α‐tocopherol, 25‐hydroxyvitamin D2, and 25‐hydroxyvitamin in plasma samples of cardiovascular patients
- Simultaneous determination of 25-hydroxyvitamin-D2 (25-OHD2), 25-hydroxyvitamin-D3 (25-OHD3), and its 3-epimer (epi-25-OHD3), retinol, and α-tocopherol in human serum samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
- Development of two ultra and high-performance liquid chromatographic methods, one coupled with atmospheric pressure chemical ionization and tandem mass spectrometric detector (UHPLC-APCI-MS/MS) and the other one with diode array detector (HPLC-DAD) to estimate fat-soluble vitamins A, D3 and E in rice cereal-based baby food samples
- Multi-residue analysis of fat-soluble vitamins (FSVs)― retinol, 25(OH)D2, 25(OH)D3, epi-25(OH)D3. and α-tocopherol in plasma and amniotic fluid samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) following a single pre-treatment step
Features and Benefits
- Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
- Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
- Rigorously tested through real-time stability studies to ensure accuracy and shelf life
- Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
- Flame sealed under argon into ampoules for long-term shelf life
- Offered in a convenient, DEA-exempt format to improve laboratory efficiency
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| application(s) clinical testing | application(s) clinical testing | application(s) clinical testing | application(s) - |
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) - |
| form liquid | form liquid | form liquid | form powder |
| format single component solution | format single component solution | format single component solution | format - |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 200 |
| grade certified reference material | grade certified reference material | grade certified reference material | grade - |
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| V-010-1ML | 04061837428524 |






