Skip to Content
MilliporeSigma

Skip To

L-032

Lacosamide-13C, D3 solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
13CC12D3H15N2O3
Molecular Weight:
254.30
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H][13C]([2H])([2H])OC[C@@H](NC(C)=O)C(=O)NCc1ccccc1

InChI

1S/C13H18N2O3/c1-10(16)15-12(9-18-2)13(17)14-8-11-6-4-3-5-7-11/h3-7,12H,8-9H2,1-2H3,(H,14,17)(H,15,16)/t12-/m1/s1/i2+1D3

InChI key

VPPJLAIAVCUEMN-RUXIMPAMSA-N

General description

An internal standard suitable for GC/MS or LC/MS applications in lacosamide testing from clinical toxicology and forensic analysis to urine drug testing and isotope dilution methods. Lacosamide is a new antiepileptic drug approved by the FDA in late 2008 for the treatment of partial-onset seizures and diabetic neuropathic pain. The drug is marketed under the trade name Vimpat®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vimpat is a registered trademark of Harris FRC Corporation

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L-027L-029B-063
technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under Lacosamide-13C, D3 solution


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
L-032-1ML04061833945421

Questions

Reviews

No rating value

Active Filters