Skip to Content
MilliporeSigma

Skip To

488682

Deuterium chloride

99 atom % D

Synonym(s):

Deutero-hydrochloric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
DCl
CAS Number:
Molecular Weight:
37.47
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12352003
EC Number:
231-715-8
MDL number:
Isotopic purity:
99 atom % D
Mass shift:
M+1
Form:
gas
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

1.3 (vs air)

Quality Segment

vapor pressure

613 psi ( 21.1 °C)

isotopic purity

99 atom % D

form

gas

bp

−85 °C (lit.)

mp

−114 °C (lit.)

mass shift

M+1

SMILES string

Cl[2H]

InChI

1S/ClH/h1H/i/hD

InChI key

VEXZGXHMUGYJMC-DYCDLGHISA-N

General description

Deuterium chloride 99 atom % D is a high-purity isotope product with a unique chemical profile that belongs to the class of isotopes. Produced by Sigma-Aldrich, we are committed to providing quality isotope products and a reliable supply chain. Available in industrial and pre-pack quantities.

Application

- Quality control reference standards
- Chemical synthesis of labeled compounds
- Analytical method development
- reaction mechanism investigations

Features and Benefits

Features
- High isotopic purity for reliable results
- Stable under various experimental conditions
- Compatible with a wide range of chemical processes
- Provides accurate insights in complex systems

Benefits
- Facilitates Advanced research methodologies and applications
- Provides reliable data for environmental monitoring
- Enhances understanding of chemical interactions
- Supports the development of innovative solutions

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Shipping of this product by air freight is forbidden.
Outside of the U.S., ocean freight shipment is required and will result in substantial additional expense.
Standard 5 L quantity is packaged in a 450 mL carbon steel lecture bottle with stainless steel CGA 110/180 valve. Nominal gas pressure at 21ºC is 150 psig for 5 L. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Monel control valve Z146978 or Monel gas regulator Z562483 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
543047532991615390
isotopic purity

99 atom % D

isotopic purity

≥99 atom % D

isotopic purity

97.5 atom % D

isotopic purity

98 atom % D

mass shift

M+1

mass shift

M+1

mass shift

M+1

mass shift

M+6

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-

form

gas

form

liquid

form

-

form

solid

bp

−85 °C (lit.)

bp

-

bp

-

bp

-

mp

−114 °C (lit.)

mp

-

mp

-

mp

114-117 °C


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Press. Gas Compr. Gas - Skin Corr. 1A

Storage Class

2A - Gases

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters