Skip to Content
Merck

Skip To

1720407

USP

Xylazine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H16N2S
CAS Number:
Molecular Weight:
220.33
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

xylazine

manufacturer/tradename

USP

drug control

Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

S1CCCN\C\1=N/c2c(cccc2C)C

InChI

1S/C12H16N2S/c1-9-5-3-6-10(2)11(9)14-12-13-7-4-8-15-12/h3,5-6H,4,7-8H2,1-2H3,(H,13,14)

InChI key

BPICBUSOMSTKRF-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Xylazine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
X1126PHR32631720429
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

format

neat

format

-

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

API family

xylazine

API family

-

API family

xylazine

API family

xylazine

drug control

Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

-


Still not finding the right product?

Explore all of our products under Xylazine


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1720407-200MG04061833834923

Questions

  1. What is the solubility of 1356009, Ketamine hydrochloride and 1720407, Xylazine?

    1 answer
    1. The solubility of 1720407, Xylazine, are as follows: DMF: 30 mg/ml; DMSO: 30 mg/ml; DMSO:PBS (pH 7.2) (1:1): 0.5 mg/ml; Ethanol: 20 mg/ml.

      For 1356009, USP, Ketamine hydrochloride, it is soluble in organic solvents such as ethanol, DMSO, and dimethyl formamide (DMF) with the following solubility: DMF: 5 mg/ml; DMSO: 10 mg/ml; Ethanol: 10 mg/ml; PBS (pH 7.2): 5 mg/ml.

      Helpful?

Reviews

No rating value

Active Filters