Skip to Content
Merck

Skip To

1711202

USP

Verapamil hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-Verapamil hydrochloride, 5-[N-(3,4-Dimethoxyphenylethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3O)2C6H3CH2CH2N(CH3)(CH2)3C[C6H3(OCH3)2][CH(CH3)2]CN · HCl
CAS Number:
Molecular Weight:
491.06
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
205-800-5
Beilstein/REAXYS Number:
3647093
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

verapamil

manufacturer/tradename

USP

mp

142 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.COc1ccc(CCN(C)CCCC(C#N)(C(C)C)c2ccc(OC)c(OC)c2)cc1OC

InChI

1S/C27H38N2O4.ClH/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6;/h9-12,17-18,20H,8,13-16H2,1-7H3;1H

InChI key

DOQPXTMNIUCOSY-UHFFFAOYSA-N

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Verapamil hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Verapamil Hydrochloride Injection
  • Verapamil Hydrochloride Tablets
  • Verapamil Hydrochloride Extended-Release Capsules
  • Verapamil Hydrochloride Extended-Release Tablets
  • Verapamil Hydrochloride Compounded Oral Solution
  • Verapamil Hydrochloride Compounded Oral Suspension

Biochem/physiol Actions

α1-adrenoceptor antagonist; L-type calcium channel blocker.
α1-adrenoceptor antagonist; L-type calcium channel blocker. Blocks L-type Ca2+ channels in smooth and cardiac muscle, induces apoptosis of human primary and metastatic colon adenocarcinoma cells in vitro. Drug resistance reversal agent acting on Pgp, e.g. decrease renal tubule elimination of digoxin. Increases basal ATPase activity of Pgp. Substrate of Cyp3A4 and CYP2C6.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V0100000BP7201205003
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

mp

142 °C (dec.) (lit.)

mp

142 °C (dec.) (lit.)

mp

142 °C (dec.) (lit.)

mp

-

API family

verapamil

API family

verapamil

API family

verapamil

API family

diltiazem


Still not finding the right product?

Explore all of our products under Verapamil hydrochloride


pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



S Izdes et al.
Acta clinica Belgica, 69(2), 116-119 (2014-04-15)
Verapamil intoxication is a life-threatening condition that often presents with severe hemodynamic instability and requires vasopressor support. There are also documented case reports of the development of non-cardiogenic pulmonary oedema after verapamil overdose. However, the exact mechanisms responsible for pulmonary
S H Baky et al.
Pharmacotherapy, 2(6), 328-353 (1982-11-01)
Verapamil hydrochloride, a prototype calcium antagonist, is now marketed in the United States for the acute treatment of supraventricular tachyarrhythmias and for chronic management of vasospastic and chronic stable angina. It inhibits the slow inward channel in in the heart
Soon Hyung Park et al.
The Laryngoscope, 124(6), 1444-1451 (2013-10-25)
We investigated whether the round window membrane (RWM) vibration can facilitate dexamethasone perfusion via the RWM in patients with sudden hearing loss. Prospective study. We first performed an in vitro study using a semipermeable membrane. In the subsequent in vivo



Global Trade Item Number

SKUGTIN
1711202-200MG04061838750693

Questions

Reviews

No rating value

Active Filters