Skip to Content
Merck

Skip To

1632004

USP

Sulfanilamide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

p-Aminobenzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
H2NC6H4SO2NH2
CAS Number:
Molecular Weight:
172.20
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
511852
eCl@ss:
39093202
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

sulfanilamide

manufacturer/tradename

USP

mp

164-166 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(cc1)S(N)(=O)=O

InChI

1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)

InChI key

FDDDEECHVMSUSB-UHFFFAOYSA-N

General description

Sulfanilamide is an antibacterial drug.

Application

Sulfanilamide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Silver Sulfadiazine
  • Sulfacetamide Sodium
  • Sulfamethoxazole
  • Sulfamethoxazole and Trimethoprim Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S21510001.1179946874
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

analytical standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

clinical testing

API family

sulfanilamide

API family

sulfanilamide

API family

-

API family

-

mp

164-166 °C (lit.)

mp

164-166 °C (lit.)

mp

162.8 °C

mp

164-166 °C (lit.)


Still not finding the right product?

Explore all of our products under Sulfanilamide


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



SPECTROSCOPIC STUDIES OF CUTANEOUS PHOTOSENSITIZING AGENTS?II. SPIN TRAPPING OF PHOTOLYSIS PRODUCTS FROM SULFANILAMIDE AND 4-AMINOBENZOIC ACID USING 5, 5-DIMETHYL-1-PYRROLINE-1-OXIDE.
Chignell, Colin F., et al.
Photochemistry and Photobiology, 34.2 , 147-156 (1981)
Mei-Hsiu Chen et al.
Lasers in medical science, 30(1), 263-272 (2014-09-19)
For decades, low-level laser therapy (LLLT) has widespread applications in tendon-related injuries. Although the therapeutic effect of LLLT could be explained by photostimulation of target tissue and cells, how tenocytes sense photonic energy and convert them into cascades of cellular
Danielle D Carrade Holt et al.
Stem cells and development, 23(11), 1258-1265 (2014-01-21)
Mesenchymal stem cells (MSCs) are used in both human clinical trials and veterinary medicine for the treatment of inflammatory and immune-mediated diseases. MSCs modulate inflammation by decreasing the cells and products of the inflammatory response. Stimulated equine MSCs from bone



Global Trade Item Number

SKUGTIN
1632004-5G04061838747471

Questions

Reviews

No rating value

Active Filters