Skip to Content
Merck

Skip To

1605952

USP

Rosmarinic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(R)-O-(3,4-Dihydroxycinnamoyl)-3-(3,4- dihydroxyphenyl)lactic acid, 3,4-Dihydroxycinnamic acid (R)-1-carboxy-2-(3,4-dihydroxyphenyl)ethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H16O8
CAS Number:
Molecular Weight:
360.31
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2227587
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

rosmarinic acid

manufacturer/tradename

USP

mp

171-175 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](Cc1ccc(O)c(O)c1)OC(=O)\C=C\c2ccc(O)c(O)c2

InChI

1S/C18H16O8/c19-12-4-1-10(7-14(12)21)3-6-17(23)26-16(18(24)25)9-11-2-5-13(20)15(22)8-11/h1-8,16,19-22H,9H2,(H,24,25)/b6-3+/t16-/m1/s1

InChI key

DOUMFZQKYFQNTF-WUTVXBCWSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Rosmarinic acid has shown to contain antioxidant and anti-inflammatory activities. Possesses promising physiological actions related to cognitive performance, Alzheimer′s disease prevention, kidney disease treatment, cardioprotection and cancer chemoprevention.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
00390580R40331609818
manufacturer/tradename

USP

manufacturer/tradename

HWI

manufacturer/tradename

-

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

primary reference standard

grade

-

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

-

format

neat

mp

171-175 °C (lit.)

mp

171-175 °C (lit.)

mp

171-175 °C (lit.)

mp

-

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under Rosmarinic acid


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1605952-25MG04061833832806

Questions

Reviews

No rating value

Active Filters