Skip to Content
Merck

Skip To

1604530

USP

Rivaroxaban

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-Rivaroxaban, 5-Chloro-N-[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-2-thiophenecarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H18ClN3O5S
CAS Number:
Molecular Weight:
435.88
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

rivaroxaban

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[s]1c(ccc1C(=O)NC[C@@H]2OC(=O)N(C2)c3ccc(cc3)N4CCOCC4=O)Cl

InChI

1S/C19H18ClN3O5S/c20-16-6-5-15(29-16)18(25)21-9-14-10-23(19(26)28-14)13-3-1-12(2-4-13)22-7-8-27-11-17(22)24/h1-6,14H,7-11H2,(H,21,25)/t14-/m0/s1

InChI key

KGFYHTZWPPHNLQ-AWEZNQCLSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Rivaroxaban USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Rivaroxaban Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0002187SML28441604541
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

-

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

-

format

neat

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

API family

rivaroxaban

API family

rivaroxaban

API family

-

API family

rivaroxaban


Still not finding the right product?

Explore all of our products under Rivaroxaban


pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1604530-200MG04061842579495

Questions

Reviews

No rating value

Active Filters