Skip to Content
Merck

Skip To

1547925

USP

Polysorbate 20

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

polysorbate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O1C(C(C(C1)OCCO)OCCO)C(OCCO)COCCOC(=O)CCCCCCCCCCC

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.Polysorbate 20 is a laurate ester of sorbitol and its anhydrides, copolymerized with approximately 20 moles of ethylene oxide for each mole of sorbitol and sorbitol anhydrides. The fatty acids may be of vegetable, animal, or synthetic origin.

Application

It is intended to be used as a USP reference standard in specified quality tests and assays.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR378944112Y0002464
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

pharmaceutical primary standard

format

neat

format

-

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

research use (small molecule)

application(s)

pharmaceutical

API family

polysorbate

API family

-

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Polysorbate 20


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

527.0 °F - Pensky-Martens closed cup

flash_point_c

275 °C - Pensky-Martens closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Polysorbate 20
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(2), 5965-5965 (2020)



Global Trade Item Number

SKUGTIN
1547925-2G04061833861059

Questions

Reviews

No rating value

Active Filters