Skip to Content
Merck

Skip To

1491332

USP

Paclitaxel

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C47H51NO14
CAS Number:
Molecular Weight:
853.91
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1420457
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

paclitaxel

manufacturer/tradename

USP

mp

213 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](O)[C@@]3(C)C(=O)[C@H](OC(C)=O)C4=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)c5ccccc5)[C@]3([H])[C@@]1(CO2)OC(C)=O)C4(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccccc7

InChI

1S/C47H51NO14/c1-25-31(60-43(56)36(52)35(28-16-10-7-11-17-28)48-41(54)29-18-12-8-13-19-29)23-47(57)40(61-42(55)30-20-14-9-15-21-30)38-45(6,32(51)22-33-46(38,24-58-33)62-27(3)50)39(53)37(59-26(2)49)34(25)44(47,4)5/h7-21,31-33,35-38,40,51-52,57H,22-24H2,1-6H3,(H,48,54)/t31-,32-,33+,35-,36+,37+,38-,40-,45+,46-,47+/m0/s1

InChI key

RCINICONZNJXQF-MZXODVADSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Paclitaxel USP reference standard meant for use in specified quality tests and assays. Also used to prepare standard and system suitability solutions for the assay and impurity analysis using the liquid chromatography method coupled to a UV detector according to the given below monographs of United States Pharmacopeia (USP):

  • Paclitaxel Injection
  • Paclitaxel

Biochem/physiol Actions

Paclitaxel is a potent anti-neoplastic and anti-mitotic taxane drug, which binds to the N-terminus of β-tubulin and and stabilizes microtubules arresting the cell cycle at the G2/M phase.
Paclitaxel is a potent anti-neoplastic and anti-mitotic taxane drug, which binds to the N-terminus of β-tubulin and and stabilizes microtubules arresting the cell cycle at the G2/M phase. The microtubule damage induces apoptosis through a JNK-dependent pathway followed by a JNK-independent pathway, perhaps related to the activation of protein kinase A (PKA) or of Raf-1 kinase, resulting in phosphorylation of Bcl-2. A major metabolite via CYP2CB is 6α-hydroxypaclitaxel (6α-OHP).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Disclaimer

Paclitaxel undergoes transesterification in methanol and hydrolyzes in aqueous solutions.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0000698Y0000739Y0000719
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

EDQM

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

mp

213 °C (dec.) (lit.)

mp

213 °C (dec.) (lit.)

mp

213 °C (dec.) (lit.)

mp

213 °C (dec.) (lit.)

API family

paclitaxel

API family

paclitaxel

API family

paclitaxel

API family

paclitaxel


Still not finding the right product?

Explore all of our products under Paclitaxel


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Muta. 2 - Repr. 1B - STOT RE 1

target_organs

Central nervous system,Bone marrow,Cardio-vascular system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters