Skip to Content
Merck

Skip To

1444707

USP

Mitomycin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Mitomycin C, Ametycine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

mitomycin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N21[C@]([C@H]4N[C@H]4C2)([C@@H](C3=C1C(=O)C(=C(C3=O)N)C)COC(=O)N)OC

InChI

1S/C15H18N4O5/c1-5-9(16)12(21)8-6(4-24-14(17)22)15(23-2)13-7(18-13)3-19(15)10(8)11(5)20/h6-7,13,18H,3-4,16H2,1-2H3,(H2,17,22)/t6-,7+,13+,15-/m1/s1

InChI key

NWIBSHFKIJFRCO-WUDYKRTCSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mitomycin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Mitomycin for Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y000037810107409001M0503
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

Roche

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

format

neat

format

neat

format

-

format

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

API family

mitomycin

API family

mitomycin

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Mitomycin


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1444707-50MG04061833845851

Questions

  1. What's the difference between mitomycin: 1444707 (USP standard) and Y0000378 (EP standard)? Why the price difference is so large?

    1 answer
    1. These products are the same compound with the same CAS number, however, they are issued by different agencies. Item 1444707 is issued by the United States Pharmacopoeia and the European Pharmacopoeia issues item Y0000378. The suppliers set the price, and the factors that influence this pricing are unknown. However, in many cases, testing and stringent specifications may play a significant role.
      Please see the links below for a sample Certificate of Analysis for each product:
      1444707:
      https://static.usp.org/pdf/EN/referenceStandards/certificates/1444707-R137C0.pdf
      Y0000378:
      https://crs.edqm.eu/db/4DCGI/leaflet?leaflet=Y0000378_3.pdf

      Helpful?

Reviews

No rating value

Active Filters