Skip to Content
Merck

Skip To

1370000

USP

Loperamide hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutyramide hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C29H33ClN2O2 · HCl
CAS Number:
Molecular Weight:
513.50
MDL number:
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

loperamide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN(C)C(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4

InChI

1S/C29H33ClN2O2.ClH/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32-20-17-28(34,18-21-32)23-13-15-26(30)16-14-23;/h3-16,34H,17-22H2,1-2H3;1H

InChI key

PGYPOBZJRVSMDS-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Application

Loperamide hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Loperamide Hydrochloride Tablets
  • Loperamide Hydrochloride Capsules
  • Loperamide Hydrochloride Oral Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1162BP63534014
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

analytical standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

BP

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

Gene Information

human ... OPRM1(4988)

Gene Information

human ... OPRM1(4988)

Gene Information

human ... OPRM1(4988)

Gene Information

human ... OPRM1(4988)

API family

loperamide

API family

loperamide

API family

loperamide

API family

-


Still not finding the right product?

Explore all of our products under Loperamide hydrochloride


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



H Miyazaki et al.
European journal of drug metabolism and pharmacokinetics, 4(4), 199-206 (1979-01-01)
Following oral administration of [14C]loperamide hydrochloride in 1 mg/kg to rats, plasma levels of radioactivity reached maximum at 4 hrs and decreased with a half-life of 4.1 hrs. Radioactivity in 96-hr feces accounted for 95% of the dose, with 30%
Loperamide hydrochloride.
L M Daugherty
American pharmacy, NS30(12), 45-48 (1990-12-01)
Jae-Suk Choi et al.
Evidence-based complementary and alternative medicine : eCAM, 2014, 878503-878503 (2014-09-13)
Aim. The objective was to evaluate the synergistic effects of fermented rice extracts (FRe) on the laxative and probiotic properties of yoghurt in rats with loperamide-induced constipation. Methods. After constipation induction, yoghurt containing FRe (BFRe; 0.05%, 0.1%, or 1%) was



Global Trade Item Number

SKUGTIN
1370000-200MG04061838731579

Questions

Reviews

No rating value

Active Filters