Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
dexamethasone
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[Na+].[Na+].[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@@H](C)[C@]4(O)C(=O)COP([O-])([O-])=O
InChI
1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15+,16+,17+,19+,20+,21+,22+;;/m1../s1
InChI key
PLCQGRYPOISRTQ-FCJDYXGNSA-L
Gene Information
human ... NR3C1(2908)
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename BP | manufacturer/tradename - | manufacturer/tradename EDQM |
| format neat | format neat | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| API family dexamethasone | API family dexamethasone | API family dexamethasone | API family dexamethasone |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. - |
Still not finding the right product?
Explore all of our products under Dexamethasone sodium phosphate
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Repr. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1177032-500MG | 04061835345991 |


