Skip to Content
Merck

Skip To

1081829

USP

2-Butanol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

sec-Butyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3CH2CH(OH)CH3
CAS Number:
Molecular Weight:
74.12
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
773649
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

2.6 (vs air)

vapor pressure

12.5 mmHg ( 20 °C)

expl. lim.

9.8 %

manufacturer/tradename

USP

refractive index

n20/D 1.397 (lit.)

bp

98 °C (lit.)

mp

−115 °C (lit.)

density

0.808 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCC(C)O

InChI

1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3

InChI key

BTANRVKWQNVYAZ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

2-Butanol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Butyl Alcohol
  • Isobutyl Alcohol
  • Pectin

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1563968701.09630
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

analytical standard

grade

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

application(s)

-

vapor density

2.6 (vs air)

vapor density

2.6 (vs air)

vapor density

2.6 (vs air)

vapor density

-

expl. lim.

9.8 %

expl. lim.

9.8 %

expl. lim.

9.8 %

expl. lim.

1.4-9.8 % (v/v)


Still not finding the right product?

Explore all of our products under 2-Butanol


pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1081829-3X1.2ML04061838679468

Questions

Reviews

No rating value

Active Filters