Skip to Content
Merck

Skip To

1029909

USP

Ammonio methacrylate copolymer type A

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

ammonio methacrylate copolymer, copolymers

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Cl-].[N+](CCOC(=O)C(=C)C)(C)(C)C.O(CC)C(=O)C=C.O(C)C(=O)C(=C)C

InChI

1S/C9H18NO2.2C5H8O2.ClH/c1-8(2)9(11)12-7-6-10(3,4)5;1-4(2)5(6)7-3;1-3-5(6)7-4-2;/h1,6-7H2,2-5H3;1H2,2-3H3;3H,1,4H2,2H3;1H/q+1;;;/p-1

InChI key

FSXVSUSRJXIJHB-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ammonio methacrylate copolymer type A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ammonio Methacrylate Copolymer
  • Ammonio Methacrylate Copolymer Dispersion

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
102991010256021396400
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

ammonio methacrylate copolymer, copolymers

API family

ammonio methacrylate copolymer, copolymers

API family

amino methacrylate copolymer, basic butylated methacrylate copolymer, copolymers

API family

copolymers, methacrylic acid copolymers


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1029909-500MG04061833839515
1029909-100MG04065272692205

Questions

Reviews

No rating value

Active Filters