Skip to Content
Merck

Skip To

1005706

USP

Glacial acetic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Acetic acid, Ethanoic acid, Methanecarboxylic acid, Glacial acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
506007
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

vapor density

2.07 (vs air)

API family

glacial acetic acid

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F, 4 %, 59 °F

manufacturer/tradename

USP

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

General description

Glacial acetic acid is a commonly used solvent and pH modifier in peptide synthesis and purification. It may be present as a residual process reagent. The USP standard is used to quantify acetic acid levels in peptide APIs and assess formulation stability.

The USP biologics peptides category encompasses a diverse range of therapeutic peptides that are essential in managing various medical conditions. These peptides, typically consisting of amino acid sequences of 40 residues or less, are critical for the development of high-quality medicines. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of peptide therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Glacial acetic acid USP reference standard for use in specified quality tests and assays. It is also used to prepare standard solution by using liquid chromatography coupled to a UV detector according to the general chapter of United States Pharmacopeia (USP). Based directly on official monographs published in the USP-NF, these highly characterized physical specimens are used in testing by pharmaceutical and related industries to help ensure the identity, strength, quality, and purity of peptide-based medicines.

Further information is available in the general chapter, 503- ACETIC ACID IN PEPTIDES, USP43-NF38 - 6741 of the USP (United States Pharmacopeia).

Other Notes

Sales restrictions may apply.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1748AX00731.37000
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

ACS reagent

grade

-

form

liquid

form

-

form

liquid

form

liquid

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

format

neat

format

neat

format

-

format

-

mp

16.2 °C (lit.)

mp

16.2 °C (lit.)

mp

16.2 °C (lit.)

mp

16.2 °C (lit.)


Still not finding the right product?

Explore all of our products under Glacial acetic acid


pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

flash_point_f

102.2 °F - closed cup

flash_point_c

39 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Glacial Acetic Acid Adverse Events: Case Reports and Review of the Literature
Doles W, et al.
Hospital pharmacy (2015)
ACETIC ACID IN PEPTIDES
USP43-NF38 - 6741:United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(5) (2017)
Acetic acid in peptides
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(5), 6741-6741 (2017)



Global Trade Item Number

SKUGTIN
1005706-3X1.5ML04061838663818

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters