Skip to Content
Merck

Skip To

T7205

Tegafur

powder

Synonym(s):

5-Fluoro-1-(tetrahydro-2-furyl)uracil, Ftorafur, Tegafur

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H9FN2O3
CAS Number:
Molecular Weight:
200.17
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
241-846-2
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to off-white

mp

171-173 °C (lit.)

solubility

DMSO: >50 mg/mL

storage temp.

room temp

SMILES string

FC1=CN(C2CCCO2)C(=O)NC1=O

InChI

1S/C8H9FN2O3/c9-5-4-11(6-2-1-3-14-6)8(13)10-7(5)12/h4,6H,1-3H2,(H,10,12,13)

InChI key

WFWLQNSHRPWKFK-UHFFFAOYSA-N

Application

Tegafur has been used as an internal standard for the analysis of 5′DFCR (5′deoxyfluorocytidine) and 5′DFUR (5′deoxyfluorouridine) using HPLC techniques[1]. Tegafur has also been used as an internal standard for the chromatographic assay of azidothymidine (AZT)[2].

Biochem/physiol Actions

Tegafur is a pro-drug of 5-fluorouracil, an antimetabolite used as an antineoplastic agent. It has been used as adjuvant chemotherapy for curatively resected colorectal cancer therapy.

Preparation Note

Tegafur is soluble in DMSO at a concentration that is greater than 50 mg/ml.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F5307PZ0215PZ0327
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

solubility

DMSO: >50 mg/mL

solubility

DMSO: >20 mg/mL

solubility

DMSO: 5 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

color

white to off-white

color

white to off-white

color

white to beige

color

white to beige


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters