Skip to Content
Merck

Skip To

T5266

Trypsin from porcine pancreas

lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.75
EC Number:
232-650-8
MDL number:
EC Number:
Specific activity:
1,000-2,000 BAEE units/mg solid
Biological source:
Porcine pancreas
Concentration:
25 mg/mL
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Porcine pancreas

Quality Segment

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

specific activity

1,000-2,000 BAEE units/mg solid

mol wt

23.8 kDa

concentration

25 mg/mL

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

Application

For trypsin digestion of peptides, use a ratio of about 1:100 to 1:20 for trypsin:peptide. The typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture. Trypsins have also been used for the re-suspension of cells during cell culture, in proteomics research for digestion of proteins and in various in-gel digestionsns†. Additional applications include assessing crystallization by membrane-based techniques and in a study to determine that protein folding rates and yields can be limited by the presence of kinetic traps.

Biochem/physiol Actions

Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. The optimal pH for trypsin activity is 7-9. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.

Serine protease inhibitors, including DFP, TLCK, APMSF, AEBSEF, and aprotinin, amongst others, will inhibit Trypsin.

Preparation Note

This product is a lyophilized powder soluble in Hank′s Balanced Salt Solution at 25 mg/mL.
For applications that involve EDTA, solubilizing trypsin should be done with a buffered salt solution contaiing no Ca2+ or Mg2+.

Other Notes

One BAEE unit will produce a ΔA253 of 0.001 per min at pH 7.6 at 25° C using BAEE as substrate. One BTEE unit = 320 ATEE units. Reaction volume = 3.2 mL (1 cm light path).
Trypsin consists of a single chain polypeptide of 223 amino acid residues, produced by the removal of the N-terminal hexapeptide from trypsinogen which is cleaved at the Lys - lle peptide bond. The sequence of amino acids is cross-linked by 6 disulfide bridges. This is the native form of trypsin, beta-trypsin. BETA-trypsin can be autolyzed, cleaving at the Lys - Ser residue, to produce alpha-trypsin. Trypsin is a member of the serine protease family.

Disclaimer

Solutions in 1 mM HCl are stable for 1 year in aliquots and stored at -20°C. The presence of Ca2+ will also diminish the self-autolysis of trypsin and maintain its stability in solution. Trypsin will also retain most of its activity in 2.0 M urea, 2.0 M guanidine HCl, or 0.1% (w/v) SDS.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T4799T0303T1426
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

technique(s)

-

technique(s)

-

specific activity

1,000-2,000 BAEE units/mg solid

specific activity

1,000-2,000 BAEE units/mg solid

specific activity

13,000-20,000 BAEE units/mg protein

specific activity

≥10,000 BAEE units/mg protein

biological source

Porcine pancreas

biological source

Porcine pancreas

biological source

-

biological source

bovine pancreas

concentration

25 mg/mL

concentration

25 mg/mL

concentration

-

concentration

-

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

essentially salt-free, lyophilized powder

sterility

γ-irradiated

sterility

-

sterility

-

sterility

-


Still not finding the right product?

Explore all of our products under Trypsin from porcine pancreas


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
T5266-500MG04061837347313

Questions

Reviews

No rating value

Active Filters