Skip to Content
Merck

Skip To

T150

(−)-Thalidomide

>98%, solid

Synonym(s):

S(−)-2-(2,6-Dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H10N2O4
CAS Number:
Molecular Weight:
258.23
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352207
MDL number:
Assay:
>98%
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


ligand

thalidomide

assay

>98%

form

solid

optical activity

[α]23/D −62.6°, c = 2 in DMF(lit.)

reaction suitability

reagent type: ligand

technique(s)

cell culture | embryo: suitable

color

white

solubility

DMSO: soluble, H2O: insoluble, ethanol: insoluble

originator

Celgene

SMILES string

O=C1CC[C@H](N2C(=O)c3ccccc3C2=O)C(=O)N1

InChI

1S/C13H10N2O4/c16-10-6-5-9(11(17)14-10)15-12(18)7-3-1-2-4-8(7)13(15)19/h1-4,9H,5-6H2,(H,14,16,17)/t9-/m0/s1

InChI key

UEJJHQNACJXSKW-VIFPVBQESA-N

Gene Information

human ... LITAF(9516), TNF(7124)
mouse ... Nos2(18126)
rat ... Nos1(24598)

General description

Thalidomide is a stereoisomer, which exists in two enantiomeric states. The S enantiomer is teratogenic. Thalidomide consists of two linked rings, a phthalimide and glutarimide ring.

Application

Thalidomide has been used to study its teratogenic effects in chicken embryos and human embryonic cells. This study reported that thalidomide causes limb defects by stabilizing PTEN, inhibiting the expression of Akt and activating caspase-dependent apoptosis[1]. Thalidomide has also been used for studying glutathione mediated teratogenic resistance in mouse embryos[2].

Biochem/physiol Actions

(-)-Thalidomide selectively inhibits the biosynthesis of tumor necrosis factor α (TNF-α), which is essential for inflammatory response. It is an anti-emetic drug and is used to treat morning sickness in pregnant women. Thalidomide is also used to treat leprosy, multiple myeloma, Crohn′s disease and human immunodeficiency virus (HIV) infection. Thalidomide also inhibits angiogenesis. It is associated with several diseases such as, peripheral neuropathy, facial palsies, Duane syndrome and autism.

Features and Benefits

This compound was developed by Celgene. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

(-)-Thalidomide is soluble in DMSO, but is insoluble in water and ethanol.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T151T1441652500
form

solid

form

powder

form

powder

form

-

assay

>98%

assay

≥98% (HPLC)

assay

≥98%

assay

-

technique(s)

cell culture | embryo: suitable

technique(s)

-

technique(s)

-

technique(s)

-

solubility

DMSO: soluble, ethanol: insoluble, H2O: insoluble

solubility

DMSO: 15 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

-

color

white

color

white to beige

color

white

color

-

reaction suitability

reagent type: ligand

reaction suitability

reagent type: ligand

reaction suitability

reagent type: ligand

reaction suitability

-


pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
T150-25MG04061833632239
T150-100MG04061837337567
T150-10MG04061833622124

Questions

Reviews

No rating value

Active Filters