Skip to Content
Merck

Skip To

SAE0152

Lysozyme from chicken egg white

free of DNA contaminants, suitable for Microbiome research, lyophilized powder, protein ≥90%, ≥39,000 units/mg protein

Synonym(s):

Lysozyme from chicken egg white, Mucopeptide N-acetylmuramoylhydrolase, Muramidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.77
EC Number:
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

chicken egg white

Quality Segment

form

powder

specific activity

≥39000 units/mg protein

mol wt

single-chain 14.3 kDa

feature

DNA free

technique(s)

cell based assay: suitable

suitability

suitable for cell lysis

application(s)

cell analysis

shipped in

ambient

storage temp.

−20°C

General description

Purified Lysozyme DNA free SAE0152 undergoes strict quality control testing to ensure the absence of detectable levels of contaminating DNA using 35 cycles PCR amplification of 16S and 18S rDNA using universal primer sets.

Lysozyme is a single chain polypeptide of 129 amino acids cross-linked with four disulfide bridges.1 It hydrolyzes b(1à 4) linkages between N-acetylmuraminic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin.2,3,4 The enzyme is often used for lysing bacterial cells by hydrolyzing the peptidoglycan present in the cell walls. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan. Gram-negative bacteria are less susceptible due to the presence of an outer membrane and a lower proportion of peptidoglycan.


The enzyme is active over a broad pH range (6.0-9.0).

Lysozyme activity: ≥39,000 units/mg protein.

For isolation of nucleic acids, lysozyme has been used in the lysis peptidoglycan layer of bacterial cell walls.


The study of microbial communities has been revolutionized in recent years by the widespread adoption of culture independent analytical techniques such as 16S rRNA gene sequencing and metagenomics. Since DNA contamination during sample preparation is a major problem of these sequence-based approaches, DNA extraction reagents free of DNA contaminates are essential. This purified lysozyme preparation (SAE0152) is purified from chicken egg white, crystallized three times, dialyzed, and supplied as a lyophilized powder. Protein content by UV absorbance is ≥ 90% with the remainder (∼10%) being buffer salts such as sodium acetate and sodium chloride, and undergoes strict quality control testing to ensure the absence of detectable levels of contaminating DNA using 35 cycles PCR amplification of 16S and 18S rDNA using universal primer sets.


Application

  • Lysozyme is used for the extraction of genomic DNA from bacterial cells.
  • Lysozyme is used as an external standard for MALDI-TOF (matrix assisted laser desorption ionization-time of flight) mass analysis.
  • Lysonzyme is used to prepare spheroplasts.
Enzyme breaks down the cell walls of bacteria; used to prepare spheroplasts.

Biochem/physiol Actions

Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan. Gram-negative bacteria are less susceptible due to the presence of an outer membrane and a lower proportion of peptidoglycan. However, these cells may be hydrolyzed in the presence of EDTA that chelates metal ions in the outer bacterial membrane.

The enzyme is active over a broad pH range (6.0 to 9.0). At pH 6.2, maximal activity is observed over a wider range of ionic strengths (0.02 to 0.100 M) than at pH 9.2 (0.01 to 0.06 M).
Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan.

Features and Benefits

Purified Lysozyme free SAE0152 undergoes strict quality control testing to ensure it will be Free of DNA contaminants, suitable for Microbiome research.

The study of microbial communities has been revolutionized in recent years by the widespread adoption of culture independent analytical techniques such as 16S rRNA gene sequencing and metagenomics. Since DNA contamination during sample preparation is a major problem of these sequence-based approaches, DNA extraction reagents free of DNA contaminates are essential.

Other Notes

One unit will lyse 0.6 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 6.2 at 25 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L3790L687662970
form

powder

form

liquid

form

powder or granules

form

powder

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

room temp

storage temp.

2-8°C

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

shipped in

ambient

shipped in

wet ice

shipped in

-

shipped in

-

feature

DNA free

feature

-

feature

-

feature

-


Still not finding the right product?

Explore all of our products under Lysozyme from chicken egg white


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Hello. I need to prepare 50mg/ml solution. What is the best solvent/buffer to use? Thank you.

    1 answer
    1. A solution of lysozyme can be prepared in DNA-free water at 10 mg/mL. See the link below for further information: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/154/458/sae0152pis.pdf

      Helpful?

Reviews

No rating value

Active Filters