Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
K58/35, monoclonal
species reactivity
mammals
concentration
~1 mg/mL
technique(s)
immunohistochemistry (frozen sections): suitable, indirect immunofluorescence: 1 μg/mL using sciatic and optic nerve sections; cultured hypocampal neurons, western blot: 1 μg/mL using crude brain membrane prep (rat)
isotype
IgG1
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN8A(6334), SCN9A(6335)
mouse ... Scn10a(20264), Scn11a(24046), Scn1a(20265), Scn3a(20269), Scn4a(110880), Scn5a(20271), Scn8a(20273), Scn9a(20274)
rat ... Scn10a(29571), Scn11a(29701), Scn1a(81574), Scn3a(497770), Scn4a(25722), Scn5a(25665), Scn8a(29710), Scn9a(78956)
General description
Immunogen
Application
- immunohistochemistry(1:50)
- immunostaining (1:200)[1]
- immunofluorescence
- immunocytochemistry (1:100)
Biochem/physiol Actions
Physical form
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source mouse | biological source rabbit | biological source mouse | biological source rabbit |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone K58/35, monoclonal | clone polyclonal | clone 4G7, monoclonal | clone polyclonal |
| Gene Information human ... SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN8A(6334), SCN9A(6335) | Gene Information human ... SCN8A(6334) | Gene Information human ... SCN8A(6334) | Gene Information human ... SCN1A(6323) |
| species reactivity mammals | species reactivity rat | species reactivity rat, mouse, human | species reactivity mouse, rat |
| antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity purified immunoglobulin |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| S8809-.1MG | 04061836961084 |



