Skip to Content
Merck

Skip To

S5504

Sigmacell Cellulose

Type 50, 50 μm

Synonym(s):

Cellulose, Cellulose powder, Cotton linters

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.56
EC Number:
232-674-9
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


type

Type 50

Quality Level

particle size

50 μm

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Application

Faster development times than Type 20 (1-4 hours) with a little loss of resolution. May yield a colored background.
High purity cellulose powders for partition chromatography.
Sigmacell cellulose is used in thin layer chromatography (TLC) and cellulose TLC adsorbents. Sigmacell cellulose has been used in studies to inform industrial applications for large-scale purification of enzymes as well as to investigate the cellulase system of Penicillium echinulatum, which has been identified as a potential cellulase producer for bioconversion processes.

Preparation Note

For TLC, simply blend a 15-20% aqueous slurry for about one minute and coat plates; dry at room temperature. Usually does not require activation before use.

Legal Information


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
310697435236C6288
Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

200

particle size

50 μm

particle size

20 μm

particle size

51 μm

particle size

(medium)

type

Type 50

type

-

type

-

type

-


Still not finding the right product?

Explore all of our products under Sigmacell Cellulose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



D W Rickey et al.
Ultrasound in medicine & biology, 21(9), 1163-1176 (1995-01-01)
Doppler ultrasound flow measurement techniques are often validated using phantoms that simulate the vasculature, surrounding tissue and blood. Many researchers use rubber tubing to mimic blood vessels because of the realistic acoustic impedance, robust physical properties and wide range of
Jue Wang et al.
Cells, 10(2) (2021-03-07)
(1) Background: It is known that sickle cells contain a higher amount of Ca2+ compared to healthy red blood cells (RBCs). The increased Ca2+ is associated with the most severe symptom of sickle cell disease (SCD), the vaso-occlusive crisis (VOC).
Annarita Ciana et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 42(3), 1139-1152 (2017-07-03)
Old human red blood cells (RBCs) have a reduced surface area with respect to young RBCs. If this decrease occurred through the release of vesicles similar to the spectrin-free vesicles that are shed in vitro under different experimental conditions or



Global Trade Item Number

SKUGTIN
S5504-1KG04061836954451
S5504-500G04061836954468

Questions

Reviews

No rating value

Active Filters