Skip to Content
Merck

Skip To

PP0500

Protease Profiler Kit

Proteases for Mass Spectrometry and Proteomics

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.56
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


quality

Proteases for Mass Spectrometry and Proteomics

storage temp.

−20°C

General description

For detailed characterization of proteins of interest, increase your protease selection with the Protease Profiler. The Protease Profiler provides four proven alternative proteases in addition to Proteomics Grade Trypsin, an enzyme solublization reagent, and an enzyme reaction buffer. Each component has been stringently purified and qualified for MALDI-MS to ensure optimal performance. Characterize with greater flexibility, perform double enzymatic digestions, and explore alternative cleavage sites with the Protease Profiler.
  • Double Enzymatic Digestion - An initial digestion may result in peptide fragments greater than 5 kDa, often above the optimum detection range for most MS systems/instruments. A second digestion using a protease with different specificity results in cleavage of the large peptides, generating smaller peptides suitable for MS.
  • Explore Alternative Proteases - Trypsin may be incompatible with some protein samples. Find the protease best suited to your protein of interest and improve the quality of your data.
  • Perform Either Solution or Gel-based Digests - Optimized protocols and reagents for both digest conditions are provided, enabling flexibility in your analysis.

Application

The Protease Profiler Kit can be used for different enzymatic digestions on the proteins of interest.

Preparation Note

The entire Protease Profiler Kit retains activity for at least 1 year when stored at –20 °C. Alternatively, some of the components (Trypsin, Lys-C, Asp-N, Trypsin Solubilization Reagent, and Enzyme Reaction Buffer) may be stored at 2–8 °C. The storage temperature indicated on the labels of these products is 2–8 °C. Storage at –20 °C will not compromise the performance of these products.

Legal Information

Protease Profiler is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
539124PP0100539125
storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

quality

Proteases for Mass Spectrometry and Proteomics

quality

-

quality

-

quality

-


Kit Components Only

Product No.
Description

  • Enzyme Solublization Reagent 1 mL

  • Enzyme Reaction Buffer 25 mL

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated 20 μg
    SDS

  • Endoproteinase Asp-N from Pseudomonas fragi mutant strain, suitable for protein sequencing, lyophilized powder 2 μg
    SDS

  • Endoproteinase Glu-C from Staphylococcus aureus V8, suitable for protein sequencing, lyophilized powder 25 μg
    SDS

  • Endoproteinase Arg-C from mouse submaxillary gland, suitable for protein sequencing, lyophilized powder 5 μg
    SDS

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters