Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.
biological source
egg yolk
Quality Level
assay
≥99% (TLC)
form
lyophilized powder
functional group
phospholipid
lipid type
phosphoglycerides
shipped in
ambient
storage temp.
−20°C
SMILES string
[Na+].OCC(O)COP([O-])(=O)OC[C@@H](O)CO
InChI
1S/C6H15O8P.Na/c7-1-5(9)3-13-15(11,12)14-4-6(10)2-8;/h5-10H,1-4H2,(H,11,12);/q;+1/p-1/t5-,6?;/m0./s1
InChI key
WYHDJJSICPMMFI-GNVLWMSISA-M
Packaging
Packaged under Argon.
1 of 1
This Item | |||
|---|---|---|---|
| functional group phospholipid | functional group phospholipid | functional group phospholipid | functional group - |
| biological source egg yolk | biological source egg yolk | biological source egg yolk (dried) | biological source egg yolk |
| assay ≥99% (TLC) | assay ≥99% (TLC) | assay - | assay ≥99% |
| form lyophilized powder | form lyophilized powder | form solid | form powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mohammad Aboulwafa et al.
Journal of bacteriology, 186(24), 8453-8462 (2004-12-04)
Plasmid-encoded His-tagged glucose permease of Escherichia coli, the enzyme IIBCGlc (IIGlc), exists in two physical forms, a membrane-integrated oligomeric form and a soluble monomeric form, which separate from each other on a gel filtration column (peaks 1 and 2, respectively).
Anna York et al.
Structure (London, England : 1993), 29(7), 731-742 (2021-03-20)
Branched Lipid II, required for the formation of indirectly crosslinked peptidoglycan, is generated by MurM, a protein essential for high-level penicillin resistance in the human pathogen Streptococcus pneumoniae. We have solved the X-ray crystal structure of Staphylococcus aureus FemX, an
Isabella Palazzo et al.
Development (Cambridge, England), 147(10) (2020-04-16)
Retinal regeneration is robust in some cold-blooded vertebrates, but this process is ineffective in warm-blooded vertebrates. Understanding the mechanisms that suppress the reprogramming of Müller glia into neurogenic progenitors is key to harnessing the regenerative potential of the retina. Inflammation
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P8318-25MG | 04061834401322 |
| P8318-100MG | 04061834401308 |
| P8318-10MG | 04061834401315 |


