Skip to Content
Merck

Skip To

P6004

Prednisolone

≥99% (HPLC), powder, glucocorticoid

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1-Dehydrocortisol, 1-Dehydrohydrocortisone, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
200-021-7
MDL number:
Beilstein/REAXYS Number:
1354103
Assay:
≥99%
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Prednisolone, ≥99%

biological source

synthetic (organic)

Quality Segment

sterility

non-sterile

assay

≥99%

form

powder

mp

240 °C (dec.) (lit.)

solubility

chloroform: methanol (1:1): 50 mg/mL, clear, colorless to faintly yellow

originator

Johnson & Johnson

shipped in

ambient

storage temp.

room temp

SMILES string

O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(O)C(CO)=O)=C1

InChI

1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

OIGNJSKKLXVSLS-VWUMJDOOSA-N

Gene Information

General description

Prednisolone prevents protease activity in vitro. It stimulates myogenesis and inhibits apoptosis. Prednisolone is used to treat Duchenne muscular dystrophy (DMD), gout arthritis and chronic obstructive pulmonary disease (COPD) exacerbations.

Application

Prednisolone has been used for the Δ1-dehydrogenation of hydrocortisone by gel entrapped arthrobacter simplex cells.

Biochem/physiol Actions

Prednisolone is a glucocorticoid with anti-inflammatory and immunosuppressive activity.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Analysis Note

Actual content given on label

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
46656PHR1043P2700000
form

powder

form

-

form

-

form

-

assay

≥99%

assay

-

assay

-

assay

-

Quality Level

200

Quality Level

100

Quality Level

300

Quality Level

-

storage temp.

room temp

storage temp.

-

storage temp.

2-30°C

storage temp.

-

solubility

chloroform: methanol (1:1): 50 mg/mL, clear, colorless to faintly yellow

solubility

-

solubility

-

solubility

-

originator

Johnson & Johnson

originator

-

originator

-

originator

-


Still not finding the right product?

Explore all of our products under Prednisolone


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters