Skip to Content
Merck

Skip To

P4850

Proteinase K from Tritirachium album

buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL

Synonym(s):

Endopeptidase K

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
eCl@ss:
32160410
Concentration:
≥10 mg/mL, ≥800 units/mL
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

form

buffered aqueous glycerol solution

mol wt

28.93 kDa

concentration

≥10 mg/mL, ≥800 units/mL

impurities

≤0.5 ppm DNA (PicoGreen® assay)

foreign activity

DNase, Nickase and RNase, none detected

storage temp.

2-8°C

Application

The product has been used to study its pre-treatment effects on the silk fibroin. The aspects analysed in this study included the crystallographic properties of hydroxyapatite (HAp), and the microstructure and microhardness of the composites. The enzyme has also been used to facilitate the access of probes to rRNA using FISH techniques to detect pathogenic Staphylococcus aureus.[1]
Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Removes endotoxins that bind to cationic proteins such as lysozyme and ribonuclease A.
Reported useful for the isolation of hepatic, yeast, and mung bean mitochondria
Determination of enzyme localization on membranes
Treatment of paraffin embedded tissue sections to expose antigen binding sites for antibody labeling.
Digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research.
Proteinase K from Tritirachium album has been used:
  • in bovine endometrial epithelial cells for herpes viral DNA extraction
  • for viral RNA extraction from nasal swabs
  • as a component of phase lock and direct PCR lysis buffer

Biochem/physiol Actions

Proteinase K has a broad specificity and degrades many proteins even in the native state. It mainly cleaves the peptide bond adjacent to the carboxyl group of aliphatic and aromatic amino acids with blocked α-amino groups.[2][3] The molecular weight of proteinase K from amino acid sequence is found to be 28,930 Da and from SDS-PAGE, it is found to be 28,500 Da.[4] The optimum pH is between 7.5-9.0[2] and its isoelectric point is 8.9.[2] Ca2+ (1-5 mM) is required for its activation. Proteinase K is inhibited by DIFP (diisopropylfluorophosphate) or PMSF (phenylmethylsulfonyl fluoride).
Proteinase K is a stable and highly reactive serine protease. Evidence from crystal and molecular structure studies indicates the enzyme belongs to the subtilisin family with an active-site catalytic triad (Asp39-His69-Ser224). It is stable in a broad range of environments: pH, buffer salts, detergents (SDS), and temperature. In the presence of 0.1-0.5% SDS, proteinase K retains activity and will digest a variety of proteins and nucleases in DNA preparations without compromising the integrity of the isolated DNA.

Physical form

Solution in 40% glycerol (v/v) containing 10 mM Tris-HCl, pH 7.5, with 1 mM calcium acetate.

Other Notes

One unit will hydrolyze urea-denatured hemoglobin to produce color equivalent to 1.0 μmole of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent).

Legal Information

PicoGreen is a registered trademark of Life Technologies

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P5568SRE0047P8044
grade

Molecular Biology

grade

-

grade

Molecular Biology

grade

-

concentration

≥10 mg/mL, ≥800 units/mL

concentration

≥10 mg/mL, ≥500 units/mL

concentration

-

concentration

-

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

lyophilized powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

mol wt

28.93 kDa

mol wt

28.93 kDa

mol wt

28,930 Da, 28.93 kDa

mol wt

28.93 kDa

foreign activity

DNase, Nickase and RNase, none detected

foreign activity

-

foreign activity

DNAse, Nickase and RNAse, none detected

foreign activity

-


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters