Skip to Content
Merck

Skip To

P4636

Poly-L-glutamic acid sodium salt

suitable for ligand binding assays, Mol wt 3,000-15,000

Synonym(s):

L-Glutamic acid homopolymer sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352209
NACRES:
NA.26
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Poly-L-glutamic acid sodium salt, mol wt 3,000-15,000

form

powder

Quality Segment

mol wt

3,000-15,000

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

N([C@@H](CCC(=O)O)C(=O)N[C@@H](CCC(=O)O)C(=O)O)C(=O)[C@@H](N)CCC(=O)O

InChI

1S/C15H23N3O10/c16-7(1-4-10(19)20)13(25)17-8(2-5-11(21)22)14(26)18-9(15(27)28)3-6-12(23)24/h7-9H,1-6,16H2,(H,17,25)(H,18,26)(H,19,20)(H,21,22)(H,23,24)(H,27,28)/t7-,8-,9-/m0/s1

InChI key

BUZMZDDKFCSKOT-CIUDSAMLSA-N

Application


  • Bulk Biopolyelectrolyte Complexes from Homopolypeptides: Solid "Salt Bridges".: Investigates the creation of solid structures from biopolyelectrolyte complexes using Poly-L-glutamic acid sodium salt, emphasizing its potential in creating innovative materials with unique properties (Digby ZA et al., 2023).

  • Polymeric Core-Shell Nanoparticles Prepared by Spontaneous Emulsification Solvent Evaporation and Functionalized by the Layer-by-Layer Method.: This research utilizes Poly-L-glutamic acid sodium salt in the production of core-shell structured nanoparticles, indicating its utility in nanoparticle functionalization and stability (Szczęch M et al., 2020).

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P1818P4761P3418
form

powder

form

powder

form

powder

form

powder

technique(s)

ligand binding assay: suitable

technique(s)

-

technique(s)

-

technique(s)

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

white to off-white

color

white to light yellow

color

white to off-white

color

white to off-white

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mol wt

3,000-15,000

mol wt

1,500-5,500 by MALLS

mol wt

15,000-50,000

mol wt

2,000-11,000


Still not finding the right product?

Explore all of our products under Poly-L-glutamic acid sodium salt


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
P4636-100MG04061832780009
P4636-500MG04061832780023
P4636-25MG04061832780016
P4636-1G04061826686232

Questions

Reviews

No rating value

Active Filters