Skip to Content
Merck

Skip To

P4265

Pepstatin A

powder, ≥75% (HPLC)

Synonym(s):

Pepsin inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
EC Number:
247-600-0
MDL number:
Beilstein/REAXYS Number:
2201362
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Pepstatin A, microbial, ≥75% (HPLC)

biological source

microbial

Quality Segment

assay

≥75% (HPLC)

form

powder

mp

233 °C (dec.) (lit.)

solubility

ethanol: acetic acid (9:1): 1 mg/mL, clear to hazy, colorless

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](C)NC(=O)C[C@H](O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)[C@@H](O)CC(O)=O

InChI

1S/C34H63N5O9/c1-17(2)12-23(37-33(47)31(21(9)10)39-34(48)30(20(7)8)38-27(42)14-19(5)6)25(40)15-28(43)35-22(11)32(46)36-24(13-18(3)4)26(41)16-29(44)45/h17-26,30-31,40-41H,12-16H2,1-11H3,(H,35,43)(H,36,46)(H,37,47)(H,38,42)(H,39,48)(H,44,45)/t22-,23-,24-,25-,26-,30-,31-/m0/s1

InChI key

FAXGPCHRFPCXOO-LXTPJMTPSA-N

Application

Pepstatin A has been used:
  • as a component of synaptoneurosome buffer to homogenize the C57BL/6 striata
  • as a component of cyclin-dependent kinase 5 (Cdk5) lysis buffer
  • as an inhibitor to determine the chymotrypsin activity of California spiny lobster

Can be used in conjunction with E64-d and Leupeptin A to inhibit the degradation of autophagic cargo inside autophagosomes. For this application, the working concentration is typically between 1-10 μM.

Biochem/physiol Actions

Pepstatin A is an inhibitor of acid proteases (aspartyl peptidases). It forms a 1:1 complex with proteases such as pepsin, renin, cathepsin D, bovine chymosin, and protease B (Aspergillus niger). The inhibitor is highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases. Solubilized γ-secretase and retroviral protease are also inhibited by Pepstatin A. It has been used to characterize proteases from several sources.
Potent inhibitor of acid proteases, including pepsin, renin and cathepsin D and many microbial aspartic proteases. Effective concentration 1 μM. Stable for about 1 day.

Preparation Note

Pepstatin can be dissolved at 1 mg/ml in 10% (v/v) acetic acid in methanol (9:1 methanol:acetic acid). The inclusion of acetic acid may be necessary to dissolve this peptide in methanol or DMSO. It has been dissolved at 1 to 2 mg/ml in ethanol, but heat may be required for complete dissolution. Solutions of Pepstatin A can be heated as high as 60 °C without any decomposition of the peptide.
Stock solutions at 1 mg/ml are stable at least a week at 4 °C. A 1 mM solution in methanol or DMSO is stable for months at -20 °C. If solutions become darker yellow, the reagent is hydrolyzing.
A working concentration of 1 μM is stable for at least one day at room temperature.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P5318A6106P7125
biological source

microbial

biological source

microbial

biological source

bovine lung

biological source

Porcine gastric mucosa

assay

≥75% (HPLC)

assay

-

assay

≥98% (SDS-PAGE)

assay

-

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

200

form

powder

form

powder

form

powder

form

powder

solubility

ethanol: acetic acid (9:1): 1 mg/mL, clear to hazy, colorless

solubility

0.8-1.2 mg/mL (1% AcOH in Methanol (mix well))

solubility

H2O: 5 mg/mL, clear to very slightly hazy

solubility

10 mM HCl: soluble 1.0 mg/mL, clear to faintly turbid, colorless

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Pepstatin A


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters