Skip to Content
Merck

Skip To

P4244

Phosphate buffered saline

liquid, pH 7.2 (25 °C), autoclaved

Synonym(s):

PBS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12161700
eCl@ss:
32129211
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphate buffered saline, Autoclaved, pH 7.2 (25 °C)

Quality Segment

description

Sodium chloride, 150 mM, and sodium phosphate, 150 mM

form

liquid

pH

7.2 (25 °C)

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

General description

Phosphate buffered saline (PBS) is an isotonic standard buffer solution. PBS imitates the pH, ion concentrations, and osmolarity of the human body. It serves as an ideal buffer for maintaining the osmolarity and physiological pH for biomedical dilutions in cell culture and protein chemistry. PBS is used for standard enzymatic tests with active metal-organic framework (MOF) biocomposites. PBS is considered non-toxic to cells.

Application

Phosphate buffered saline has been used:
  • for washing bacterial cells after harvesting by centrifugation
  • for washing the peritoneal cavity for isolation of peritoneal macrophages
  • as a control and to dilute phytohemagglutinin (PHA) to prepare subcutaneous injections and study the immune response in birds.[1][2]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P3688P3744P5368
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing
histology
sample preparation

form

liquid

form

powder

form

powder

form

powder

pH

7.2 (25 °C)

pH

7.4

pH

7.6 (25 °C)

pH

7.4

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

description

Sodium chloride, 150 mM, and sodium phosphate, 150 mM

description

-

description

-

description

-


Still not finding the right product?

Explore all of our products under Phosphate buffered saline


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters