Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
CAS Number:
UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
232-642-4
NACRES:
NA.54
MDL number:
Specific activity:
≥3 units/mg solid
Biological source:
Aspergillus sp. (A. melleus)
Pricing and availability is not currently available.
biological source
Aspergillus sp. (A. melleus)
Quality Segment
type
Type XXIII
form
solid
specific activity
≥3 units/mg solid
storage temp.
2-8°C
Application
Proteinase is an enzyme used to break down proteins by hydrolyzing peptide bonds. Proteinase is used to degrade proteins, to study proteinase inhibitors and to study thermal inactivation kinetics. Proteinase is used in nucleic acid isolation procedures in incubations. It is used to study proteinase-activated receptors, such as the transducers of proteinase-mediated signaling in inflammation and the immune response[1]. Product P4032 is from Aspergillus melleus and has been used to non-specifically degraded xylanase from Streptomyces halstedii[2].
Biochem/physiol Actions
Proteinase catabolizes proteins by hydrolysis of peptide bonds. Proteases are inactivated by serine active-site inhibitors, such as phenylmethylsulfonyl fluoride (PMSF) and diisopropylfluorophosphate .
Other Notes
One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent), unless otherwise indicated.
1 of 1
This Item | |||
|---|---|---|---|
| specific activity ≥3 units/mg solid | specific activity ~100 U/mg | specific activity ≤0.05 Kunitz units/mg solid RNase, ≤5.0 Kunitz units/mg solid DNase, 7.0-14.0 units/mg solid | specific activity ≥2.4 U/g |
| biological source Aspergillus sp. (A. melleus) | biological source Aspergillus sp. (A. oryzae) | biological source - | biological source - |
| form solid | form powder | form lyophilized powder | form aqueous solution |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
| Quality Level 200 | Quality Level 200 | Quality Level 300 | Quality Level 200 |
| type Type XXIII | type - | type Type XXIV | type - |
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
QC Methods
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P4032-5G | 04061834373506 |
| P4032-25G | 04061834373483 |





