Skip to Content
Merck

Skip To

P1944

Phenol:Chloroform:Iso-amyl alcohol (125:24:1)

Molecular Biology

Synonym(s):

phenol chloroform RNA extraction

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.52
UNSPSC Code:
12190000
Assay:
≥99.0%
Grade:
Molecular Biology
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

assay

≥99.0%

form

liquid

pH

4.3-4.7

application(s)

life science and biopharma

storage temp.

2-8°C

General description

Phenol:Chloroform:Iso-amyl alcohol (125:24:1) is used for the purification of total RNA from mixtures of crude cell extracts and can eliminate the need for a time consuming ultracentrifugation step. The addition of phenol, chloroform, and isoamyl can be employed to aid in the separation of lipids and cellular debris, leading to the transfer of these components to the organic phase. Meanwhile, the genomic DNA (gDNA) remains in the aqueous phase, effectively isolating it from the other substances.

Application

Phenol:Chloroform:Iso-amyl alcohol (125:24:1) has been used:
  • for the extraction and purification of RNA.
  • to isolate RNA from Arabidopsis siliques.
  • to extract proteins and DNA from polysomal and non-polysomal fractions.
  • in TRIzol methods to prepare and extract miRNAs from cells, tissue, and whole flies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P3803P2069P4682
grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

assay

≥99.0%

assay

≥99.0%

assay

≥99.0%

assay

-

application(s)

life science and biopharma

application(s)

agriculture

application(s)

agriculture

application(s)

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

liquid

form

liquid

form

liquid

form

liquid

pH

4.3-4.7

pH

6.5-6.9(pH of phenolic phase)

pH

6.5-6.9(pH of phenolic phase), 7.8-8.2(pH of phenolic phase after the addition of Equilibration Buffer)

pH

4.1-4.5 (25 °C)


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Oral - STOT RE 2

target_organs

Liver,Kidney, Nervous system,Kidney,Liver,Skin

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

175.0 °F - closed cup

flash_point_c

79.44 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Molecular approaches for analysis of Drosophila MicroRNAs
Pandey M, et al.
Journal of Separation Science, 169-188 (2020)
Exploring translation regulation by global analysis of ribosomal association
Melamed D, et al.
Methods in Enzymology, 48(3), 301-305 (2009)
Autophagy controls resource allocation and protein storage accumulation in Arabidopsis seeds
Berardino JD, et al.
Journal of Experimental Botany, 69(6), 1403-1414 (2018)



Global Trade Item Number

SKUGTIN
P1944-400ML04061834357599
P1944-100ML04061834357353

Questions

  1. Hello, Your product is fractionated into two phases. Should I use lower phase for RNA extraction? Thank you

    1 answer
    1. Product P1944 - Phenol:Chloroform: Iso-amyl alcohol (125:24:1) is acidic. As such, the RNA isolates in an acidic pH and will partition the genomic DNA into the organic phase leaving only the RNA in the top aqueous phase.

      Helpful?

Reviews

No rating value

Active Filters