Skip to Content
Merck

Skip To

P0982

JumpStart REDTaq® ReadyMix Reaction Mix

for PCR

JumpStart™ REDTaq® ReadyMix™ Reaction Mix

Synonym(s):

Hot start PCR master mix, Hot start master mix, Hot start taq master mix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41106300
NACRES:
NA.55
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

usage

sufficient for 100 reactions, sufficient for 20 reactions, sufficient for 800 reactions

feature

dNTPs included, hotstart

concentration

2.5 units/reaction (50 μL reaction volume)

technique(s)

PCR: suitable

color

red

input

purified DNA

application(s)

agriculture

shipped in

wet ice

storage temp.

−20°C

General description

JumpStart REDTaq® ReadyMixPCR Reaction Mix is a prepared solution combining the performance benefits of hot start PCR with the dual convenience of Sigma′s ReadyMix and REDTaq. The Master mix includes Sigma′s JumpStart Taq DNA Polymerase, 99% pure deoxynucleotides, buffer, and an inert red dye in a 2× optimized reaction concentrate. Add 25 μL of the ReadyMix to primers, template, and water to a final reaction volume of 50 μL. JumpStart Taq antibody in the reaction mix inactivates the Taq DNA polymerase at room temperature. During the first denaturation step of PCR the complex dissociates and polymerase activity is fully restored. The inert red dye does not effect automated sequencing, restriction enzyme digestion, ligation, or other downstream manipulations. The PCR product is easily separated from the dye by standard purification methods if desired. The hot start mechanism allows for room temperature set up, making it the ideal for high throughput applications.

Application

Jumpstart REDTaq® ReadyMix Reaction Mix is used for direct DNA extraction and amplification using PCR.[1][2][3][4] It has also been used for:
  • qualitative multiplex PCR[1]
  • methylation-specific PCR (MSPCR)
  • PCR amplification of cDNA synthesized from total RNA

Features and Benefits

  • JumpStart REDTaq® DNA polymerase, an antibody inactivated hot start enzyme, is designed to minimize non-specific amplification while increasing target yield & specificity
  • JumpStart Taq DNA polymerase provides superior amplification regardless of template concentration
  • REDTaq JumpStart ReadyMix reduces pipetting steps and risk of contamination. The hot start mechanism allows for room temperature set up, making it the ideal for high throughput applications
  • Inert red dye allows for easy verification that reagent has been mixed properly
  • Other loading dyes are not required. Aliquots may be directly loaded onto an agarose gel after the reaction

Packaging

Default reaction volume is 50 μL

20RXN is packaged as 1 X 500 μL
100RXN is packaged as 1 X 2.5 mL
800RXN is packaged as 1 X 20 mL

Other Notes

For a typical PCR reaction, mix 25 μL of JumpStart REDTaq® ReadyMix with 25 μL of a mixture containing template DNA, primers, and water. Reaction volume can be scaled if desired.
One unit incorporates 10 nmol of total dNTPs into acid precipitable DNA in 30 min. at 74 °C.
View more detailed information on JumpStart REDTaq and ReadyMix products at www.sigma-aldrich.com/hotstart.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.
Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: US 8,404,464 and US 7,972,828. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims.
JumpStart is a trademark of Sigma-Aldrich Co. LLC
REDTaq is a registered trademark of Merck KGaA, Darmstadt, Germany
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P1107R2523P2893
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

feature

dNTPs included, hotstart

feature

Difficult Templates/Specialty Enzymes PCR, dNTPs included, hotstart

feature

dNTPs included, hotstart: no, Standard PCR

feature

Multiplex PCR, dNTPs included, hotstart

usage

sufficient for 100 reactions, sufficient for 800 reactions, sufficient for 20 reactions

usage

sufficient for 100 reactions, sufficient for 400 reactions

usage

sufficient for 100 reactions, sufficient for 20 reactions

usage

sufficient for 100 reactions, sufficient for 400 reactions

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

application(s)

agriculture

application(s)

agriculture

application(s)

agriculture

application(s)

agriculture

concentration

2.5 units/reaction (50 μL reaction volume)

concentration

0.75 units/reaction (50 μL reaction volume)

concentration

-

concentration

2.5 units/reaction (50 μL reaction volume)


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
P0982-800RXN04061835574537
P0982-100RXN04061835574520
P0982-20RXN04061834353621

Questions

Reviews

Active Filters

  1. 1 Ratings-only review