Skip to Content
Merck

Skip To

M8631

Melanin

Synthetic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.83
EC Number:
232-473-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

storage temp.

−20°C

SMILES string

[nH]1c2c3c(c4c5c([nH]c4)c([c]([c](c5c3c1)=O)=O)C)[c]([c](c2C)=O)=O

InChI

1S/C18H10N2O4/c1-5-13-9-7(3-19-13)12-10-8(11(9)17(23)15(5)21)4-20-14(10)6(2)16(22)18(12)24/h3-4,19-20H,1-2H3

InChI key

XUMBMVFBXHLACL-UHFFFAOYSA-N

General description

Melanin is a vital pigment present on the surface structures of vertebrates, which contribute to skin color. It has three forms such as, eumelanin, pheomelanin and neuromelanin. Melanin has cation chelating and redox properties. It is a potent absorbent of light. Melanin functions as a chemoprotective and photoprotective pigment. Mutations in this gene are associated with oculocutaneous albinism, vitiligo, piebaldism and Waardenburg syndrome.

Application

Melanin has been used:
  • in spectrophotometry
  • to study its effect on reverse transcription polymerase chain reaction (RT-PCR) inhibition and effect of BSA (bovine serum albumin) in overcoming the inhibition
  • as an absorber for skin layer in near-infrared tissue spectra
  • Raman spectroscopy

Preparation Note

Prepared by oxidation of tyrosine with hydrogen peroxide.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M04188.0861452549
form

powder

form

powder

form

powder

form

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

no temp limit

storage temp.

-

Quality Level

100

Quality Level

-

Quality Level

200

Quality Level

100


Still not finding the right product?

Explore all of our products under Melanin


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
M8631-100MG04061834063360
M8631-250MG04061834063384
M8631-1G04061834063377

Questions

1–6 of 6 Questions  
  1. Can this be diluted in DPBS?

    1 answer
    1. The product's solubility is assessed by preparing a 20 mg/ml solution in 1N NH4OH. Data characterizing its solubility in other solvents is not available.

      Helpful?

  2. Is it soluble in KOH and NaOH?

    1 answer
    1. The solubility of this product is tested in 1N NH4OH. Melanin is soluble in alkaline solutions and is expected to be soluble in both KOH and NaOH solutions.

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  4. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  5. Can this product be dissolved in water?

    1 answer
    1. Product M8631 is not tested for solubility in water. The solubility is tested in 1N NH4OH at 20 mg/ml.

      Helpful?

  6. We have used this melanin product. Is it known whether it is in oxidized form? Any information about the exact chemical structure ?

    1 answer
    1. This synthetic melanin product was prepared by oxidation of tyrosine with hydrogen peroxide.

      Helpful?

Reviews

No rating value

Active Filters