Skip to Content
Merck
All Photos(5)

Documents

M1765

Sigma-Aldrich

1-Lauroyl-rac-glycerol

≥99%

Synonym(s):

1-Monododecanoyl-rac-glycerol, 1-Monolaurin, 1-Monolauroyl-rac-glycerol, rac-Glycerol 1-laurate, DL-α-Laurin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3C(CH2)10COOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
274.40
Beilstein:
1726740
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

Assay

≥99%

form

powder

functional group

ketone

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC(=O)OCC(O)CO

InChI

1S/C15H30O4/c1-2-3-4-5-6-7-8-9-10-11-15(18)19-13-14(17)12-16/h14,16-17H,2-13H2,1H3

InChI key

ARIWANIATODDMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Monolaurin (1-Lauroyl-rac-glycerol) is used as an antimicrobial agent in various formulas and microemulsions and as a methane mitigation agent in ruminants.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ketan Patel et al.
Daru : journal of Faculty of Pharmacy, Tehran University of Medical Sciences, 21(1), 27-27 (2013-03-28)
Lumefantrine, an antimalarial molecule has very low and variable bioavailability owing to its extremely poor solubility in water. It is recommended to be taken with milk to enhance its solubility and bioavailability. The aim of present study was to develop
Amlan K Patra et al.
Phytochemistry, 71(11-12), 1198-1222 (2010-06-24)
Recently, greenhouse gas emissions have been of great concern globally. Ruminant livestock due to production of methane during normal fermentation in the rumen contributes substantially to the greenhouse effects. During the recent decade, a paradigm shift has been initiated whether
H Shimada et al.
Journal of natural products, 60(4), 417-418 (1997-04-01)
Brine shrimp lethality-directed fractionation of the 95% EtOH extract of the powdered, dried berries of Serenoa repens (Bart.) Small (saw-palmetto) (Palmae) led to the isolation of two monoacylglycerides, 1-monolaurin (1) and 1-monomyristin (2). Compounds 1 and 2 showed moderate biological
Kristi L Strandberg et al.
Antimicrobial agents and chemotherapy, 54(2), 597-601 (2009-12-17)
We investigated the effects of glycerol monolaurate (GML) on Lactobacillus, Candida, and Gardnerella vaginalis human vaginal microflora. Our previous work demonstrated that 6 months of GML treatment vaginally does not alter lactobacillus counts in monkeys. Candida and G. vaginalis are
M A Al-Holy et al.
Letters in applied microbiology, 50(3), 246-251 (2009-12-23)
To investigate the effect of lactic acid (LA), copper (II), and monolaurin as natural antimicrobials against Cronobacter in infant formula. The effect of LA (0.1, 0.2 and 0.3% v/v), copper (II) (10, 50 and 100 microg ml(-1)) and monolaurin (1000

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service