Skip to Content
Merck

Skip To

M0164

MES hemisodium salt

dry powder

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid hemisodium salt, 4-Morpholineethanesulfonic acid hemisodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H13NO4S·0.5Na
CAS Number:
Molecular Weight:
206.73
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

dry powder

Quality Segment

useful pH range

5.5-6.7

pKa 

6.1

SMILES string

[Na+].OS(=O)(=O)CCN1CCOCC1.[O-]S(=O)(=O)CCN2CCOCC2

InChI

1S/2C6H13NO4S.Na/c2*8-12(9,10)6-3-7-1-4-11-5-2-7;/h2*1-6H2,(H,8,9,10);/q;;+1/p-1

InChI key

KHQSUKMBWPKXOP-UHFFFAOYSA-M

General description

MES sodium salt is commonly employed as buffer.

Application

MES hemisodium salt may be employed as buffer to investigate the two-dimensional and three-dimensional (3D) patterns within multilayered poly(ethylene glycol) diacrylate (PEG-DA) hydrogels.[1]

Packaging

foil pouches

Preparation Note

Contents of each pouch dissolved in 1 liter deionized water will yield 0.1 M sodium MES buffer, pH 6.1 at 25°C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M3885M3058M3671
Quality Level

200

Quality Level

400

Quality Level

400

Quality Level

400

form

dry powder

form

crystalline powder

form

crystalline powder

form

powder

useful pH range

5.5-6.7

useful pH range

5.5-6.7

useful pH range

5.5-6.7

useful pH range

5.5-6.7

pKa 

6.1

pKa 

6.1

pKa 

6.1

pKa 

6.1


Still not finding the right product?

Explore all of our products under MES hemisodium salt


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters